Skip to Content
Merck
All Photos(3)

Key Documents

275905

Sigma-Aldrich

Tetraammineplatinum(II) chloride hydrate

98%

Synonym(s):

Tetraammineplatinum chloride hydrate, Tetraammineplatinum dichloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pt(NH3)4Cl2·xH2O
CAS Number:
Molecular Weight:
334.11 (anhydrous basis)
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

composition

Degree of hydration, ≤1

reaction suitability

core: platinum
reagent type: catalyst

SMILES string

N.N.N.N.O.Cl[Pt]Cl

InChI

1S/2ClH.4H3N.H2O.Pt/h2*1H;4*1H3;1H2;/q;;;;;;;+2/p-2

InChI key

GWBDNMCYHWRSOH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Tetraammineplatinum(II) chloride hydrate can be used as a source of platinum:
  • To deposit layer of Pt electrode on Nafion membranes.
  • To be used as a coupling catalyst in the electron beam technology for the decomposition of volatile organic compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel hybrid technology for VOC control using an electron beam and catalyst.
Jeon EC, et al.
Research on Chemical Intermediates, 34(8-9), 863-870 (2008)
Electro-mechanical actuator with muscle memory.
Khaldi A, et al.
Journal of Material Chemistry A, 2(38), 8029-8034 (2014)
Fabrication and characterization of an ionic polymer-metal composite bending sensor.
Song DS, et al.
Macromolecular Research, 25(12), 1205-1211 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service