Skip to Content
Merck
All Photos(1)

Key Documents

274372

Sigma-Aldrich

3,3-Dimethylallyl bromide

95%, contains silver wool as stabilizer

Synonym(s):

1-Bromo-3-methyl-2-butene, Prenyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2Br
CAS Number:
Molecular Weight:
149.03
Beilstein:
1420778
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

contains

silver wool as stabilizer

refractive index

n20/D 1.489 (lit.)

bp

82-83 °C/150 mmHg (lit.)

density

1.29 g/mL at 20 °C (lit.)

functional group

alkyl halide
bromo

storage temp.

2-8°C

SMILES string

C\C(C)=C\CBr

InChI

1S/C5H9Br/c1-5(2)3-4-6/h3H,4H2,1-2H3

InChI key

LOYZVRIHVZEDMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,3-Dimethylallyl bromide was used in the synthesis of 1- and 2-allylic-3-methylindoles. It was also used in the synthesis of products and base-modified pyrimidine nucleosides.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrophilic substitution in indoles?I: Model experiments related to the synthesis of echinulin.
Tetrahedron, 21(5), 989-1000 (1965)
Tetrahedron, 49, 2533-2533 (1993)
Kamal Rullah et al.
Bioorganic & medicinal chemistry letters, 24(16), 3826-3834 (2014-07-17)
The discovery of potent inhibitors of prostaglandin E2 (PGE2) synthesis in recent years has been proven to be an important game changer in pharmaceutical industry. It is known that excessive production of PGE2 triggers a vast array of biological signals
Journal of the Chemical Society. Perkin Transactions 1, 1153-1153 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service