Skip to Content
Merck
All Photos(1)

Documents

267317

Sigma-Aldrich

Rhenium

foil, thickness 0.25 mm, 99.98% trace metals basis

Synonym(s):

Rhenium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Re
CAS Number:
Molecular Weight:
186.21
MDL number:
UNSPSC Code:
12141737
PubChem Substance ID:
NACRES:
NA.23

Assay

99.98% trace metals basis

form

foil

description

19.3 μΩ-cm, 20°C

thickness

0.25 mm

bp

5596 °C (lit.)
5627 °C (lit.)

mp

3180 °C (lit.)

density

21.02 g/cm3 (lit.)

SMILES string

[ReH]

InChI

1S/Re

InChI key

WUAPFZMCVAUBPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rhenium is a very hard, corrosion, and wear-resistant rare metal with ahigh atomic number. It is widely used in the field of catalysis and the preparationof alloys for jet engines.

Application

Rhenium can be used as a working electrode for electrochemical hydrogen evolution reactions.

It can be used as an additive to prepare a molybdenum-titanium-zirconium (TZM) alloy joint to improve its tensile strength.

It can also be used as a catalyst for various hydrodeoxygenation reactions.

Quantity

3.3 g = 25 × 25 mm; 13.2 g = 50 × 50 mm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cinzia Spagnul et al.
Journal of inorganic biochemistry, 122, 57-65 (2013-03-12)
The synthesis and characterization of two novel water soluble porphyrins with three meso pyridyl rings and one peripheral chelator - either a diethylenetriamine unit (4) or a bipyridyl fragment (8) - for binding to the {(99m)Tc(CO)3}(+) moiety is reported. In
Shalina C Bottorff et al.
Inorganic chemistry, 52(6), 2939-2950 (2013-03-06)
The viability of the Huisgen cycloaddition reaction for clickable radiopharmaceutical probes was explored with an alkyne-functionalized 2-[(pyridin-2-ylmethyl)amino]acetic acid (PMAA) ligand system, 3, and fac-[M(I)(OH2)3(CO)3](+) (M = Re, (99m)Tc). Two synthetic strategies, (1) click, then chelate and (2) chelate, then click
Yanfeng Jiang et al.
Journal of the American Chemical Society, 135(10), 4088-4102 (2013-02-07)
Diiodo Re(I) complexes [ReI2(NO)(PR3)2(L)] (3, L = H2O; 4 , L = H2; R = iPr a, Cy b) were prepared and found to exhibit in the presence of "hydrosilane/B(C6F5)3" co-catalytic systems excellent activities and longevities in the hydrogenation of
Jan Dietrich et al.
Inorganic chemistry, 52(3), 1248-1264 (2013-01-15)
New cationic metallo ligands L1-L3 based on bis(terpyridine) ruthenium(II) complexes decorated with differently substituted 2,2'-bipyridines attached via amide groups (5-NHCO-bpy, 4-CONH-bpy, 5-CONH-bpy) were prepared. Coordination of Re(I)Cl(CO)(3) fragments to the bpy unit gives the corresponding bimetallic Ru~Re complexes 1-3. Hydrogen
Theshini Perera et al.
Inorganic chemistry, 52(5), 2412-2421 (2013-02-21)
We demonstrate that a tertiary sulfonamide group, N(SO2R)R'2, can rehybridize to form a M-N bond of normal length even when the group is in a linear tridentate ligand, such as in the new tridentate N(SO2R)dpa ligands derived from di-(2-picolyl)amine (N(H)dpa).

Articles

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service