Skip to Content
Merck
All Photos(1)

Documents

267163

Sigma-Aldrich

Platinum

wire, diam. 0.127 mm, 99.99% trace metals basis

Synonym(s):

Platinum element, Platinum gray

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
EC Number:
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

wire

resistivity

10.6 μΩ-cm, 20°C

diam.

0.127 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Application

Platinum wire may be used as an electrode during various electrochemical studies. It may also be used during the solid state deposition of colorless transparent crystals of a noncentrosymmetric hexaborate, KBOC, on it surface. Platinum wire, as a catalyst may be useful in accelerating the generation of hydrogen from Ni-rich magnesium alloy in NaCl solution.

Quantity

270 mg = 100 cm; 1.3 g = 500 cm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wire-like charge transport at near constant bridge energy through fluorene oligomers.
Goldsmith RH, et al.
Proceedings of the National Academy of Sciences of the USA, 102(10), 3540-3545 (2005)
K3B6O10Cl: a new structure analogous to perovskite with a large second harmonic generation response and deep UV absorption edge.
Wu H, et al.
Journal of the American Chemical Society, 133(20), 7786-7790 (2011)
Evaluation of a new hydrogen generating system: Ni-rich magnesium alloy catalyzed by platinum wire in sodium chloride solution.
Cho CY, et al.
Materials Transactions, 46(12), 2704-2708 (2005)
Loris De Cecco et al.
PloS one, 8(3), e58849-e58849 (2013-03-14)
We recently reported that peritumoral CpG-ODN treatment, activating TLR-9 expressing cells in tumor microenvironment, induces modulation of genes involved in DNA repair and sensitizes cancer cells to DNA-damaging cisplatin treatment. Here, we investigated whether this treatment induces modulation of miRNAs
Wei Sun et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 1907-1913 (2013-03-19)
In this paper a platinum (Pt) nanoparticle decorated graphene (GR) nanosheet was synthesized and used for the investigation on direct electrochemistry of myoglobin (Mb). By integrating GR-Pt nanocomposite with Mb on the surface of carbon ionic liquid electrode (CILE), a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service