Skip to Content
Merck
All Photos(3)

Key Documents

266132

Sigma-Aldrich

Manganese

powder, −325 mesh, ≥99% trace metals basis

Synonym(s):

Manganese element, Manganese-55

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mn
CAS Number:
Molecular Weight:
54.94
EC Number:
MDL number:
UNSPSC Code:
12141724
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99% trace metals basis

form

powder

resistivity

185 μΩ-cm, 20°C

particle size

−325 mesh

bp

1962 °C (lit.)

mp

1244 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[Mn]

InChI

1S/Mn

InChI key

PWHULOQIROXLJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D G Barceloux
Journal of toxicology. Clinical toxicology, 37(2), 293-307 (1999-06-26)
Manganese is a very hard, brittle metal, which is used to increase the strength of steel alloys. Absorption from the gastrointestinal tract occurs in the divalent and tetravalent forms. Permanganates, which are strong oxidizing agents, have a +7 valence. The
Alan Woolf et al.
Environmental health perspectives, 110(6), 613-616 (2002-06-11)
The patient's family bought a home in a suburb, but the proximity of the house to wetlands and its distance from the town water main prohibited connecting the house to town water. The family had a well drilled and they
M Baldwin et al.
Neurotoxicology, 20(2-3), 343-353 (1999-07-01)
Exposure data and bioindicators were obtained for a study whose objective was detection of early manifestations of manganese (Mn) neurotoxicity in a population with potential environmental exposure. The study included persons with no history of neurotoxic workplace exposure in Southwest
D Mergler et al.
Neurotoxicology, 20(2-3), 327-342 (1999-07-01)
Excessive manganese (Mn) has been associated with neurobehavioral deficits and neurological and/or neuropsychiatric illness, but the level at which this metal can cause adverse neurotoxic effects, particularly with long-term exposure, is still unknown. The objective of the present study was
Rachel J Service et al.
Biochemistry, 52(47), 8452-8464 (2013-10-31)
In the 1.9 Å structural model of photosystem II (PDB: 3ARC), the amino acid residue Glu333 of the D1 polypeptide coordinates to the oxygen-evolving Mn₄CaO₅ cluster. This residue appears to be highly significant in that it bridges the two Mn

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service