Skip to Content
Merck
All Photos(1)

Documents

264458

Sigma-Aldrich

3,4-Difluorobenzyl bromide

98%

Synonym(s):

α-Bromo-3,4-difluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH2Br
CAS Number:
Molecular Weight:
207.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.526 (lit.)

density

1.618 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(CBr)cc1F

InChI

1S/C7H5BrF2/c8-4-5-1-2-6(9)7(10)3-5/h1-3H,4H2

InChI key

JJIFTOPVKWDHJI-UHFFFAOYSA-N

Application

3,4-Difluorobenzyl bromide has been used in the synthesis of series of benzothiophene-based phosphonates.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jay M Matthews et al.
Bioorganic & medicinal chemistry letters, 22(8), 2922-2926 (2012-03-17)
A series of benzothiophene-based phosphonates was synthesized and many analogs within the series were shown to be potent antagonists of the TRPM8 channel. The compounds were obtained as a racemic mixture in 5 synthetic steps, and were tested for TRPM8

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service