257583
8-Bromooctanoic acid
97%
Synonym(s):
8-Bromocaprylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Br(CH2)7CO2H
CAS Number:
Molecular Weight:
223.11
Beilstein:
1756103
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
reaction suitability
reagent type: cross-linking reagent
bp
147-150 °C/2 mmHg (lit.)
mp
35-37 °C (lit.)
functional group
bromo
carboxylic acid
SMILES string
OC(CCCCCCCBr)=O
InChI
1S/C8H15BrO2/c9-7-5-3-1-2-4-6-8(10)11/h1-7H2,(H,10,11)
InChI key
BKJFDZSBZWHRNH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
8-Bromooctanoic acid, also known as 8-bromocaprylic acid, is a cross-linking reagent. It can undergo hydrolysis to form 8-hydroxyoctanoic acid.
Application
8-Bromooctanoic acid can be used as a crosslinking reagent to:
- prepare 8-mercaptooctanoic acid in the biosynthesis of lipoic acid
- attach triple helix-forming oligonucleotides (TFOs) to inhibitors in their synthesis
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Biosynthesis of lipoic acid: characterization of the lipoic acid auxotrophs Escherichia coli W1485-lip2 and JRG33-lip9
MA Hayden
Biochemistry, 32, 3778-3782 (1993)
Triple helix-forming oligonucleotides conjugated to new inhibitors of topoisomerase II: synthesis and binding properties
M Duca
Bioconjugate Chemistry, 16, 873-884 (2005)
Synthesis of heteroatom-substituted analogues of stearic acid.
RA Pascal
Journal of Lipid Research, 27, 221-224 (1986)
Soumen Das et al.
Journal of labelled compounds & radiopharmaceuticals, 61(14), 1048-1057 (2018-09-02)
123 I-Iodophenylpentadecanoic acid (IPPA) is a metabolic agent used in nuclear medicine for diagnosis of myocardial defects. Efforts are underway worldwide to develop a 99m Tc substitute of the above radiopharmaceutical for the aforementioned application. Herein, we report synthesis and
Beibei Hou et al.
Materials science & engineering. C, Materials for biological applications, 74, 94-102 (2017-03-04)
How to encapsulate and transport the payload of multiple therapeutic compounds avoiding premature leakage, and simultaneously co-release them rapidly at specific lesions still remains the major concern in clinic. Herein, we designed the UCN@mSiO
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service