Skip to Content
Merck
All Photos(1)

Key Documents

256781

Sigma-Aldrich

Diethylzinc

≥52 wt. % Zn basis

Synonym(s):

DEZn, Et2Zn, DEZ, Zincdiethyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2Zn
CAS Number:
Molecular Weight:
123.51
Beilstein:
3587207
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

≥52 wt. % Zn

refractive index

n20/D 1.498 (lit.)

bp

117 °C (lit.)

mp

−28 °C (lit.)

density

1.205 g/mL at 25 °C (lit.)

SMILES string

CC[Zn]CC

InChI

1S/2C2H5.Zn/c2*1-2;/h2*1H2,2H3;

InChI key

HQWPLXHWEZZGKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethylzinc has been used as a zinc precursor for the synthesis of colloidal Cu/Zn-based nanoparticles, and zinc on silica catalysts via atomic layer deposition (ALD) technique.

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Single-site zinc on silica catalysts for propylene hydrogenation and propane dehydrogenation: Synthesis and reactivity evaluation using an integrated atomic layer deposition-catalysis instrument
Camacho-Bunquin J, et al.
J. Catal., 345, 170-182 (2017)
Bifunctional catalysts based on colloidal Cu/Zn nanoparticles for the direct conversion of synthesis gas to dimethyl ether and hydrocarbons
Gentzen, M and Doronkin, DE and Sheppard, TL and Grunwaldt, J-D and Sauer, J and Behrens, S
Applied Catalysis A: General, 557, 99-107 (2018)
Bifunctional hybrid catalysts derived from Cu/Zn-based nanoparticles for single-step dimethyl ether synthesis
Gentzen, M and Habicht, W and Doronkin, DE and Grunwaldt, J-D and Sauer, J and Behrens, S
Catalysis Science & Technology, 6(4), 1054-1063 (2016)
Tummanapalli Satyanarayana et al.
Organic letters, 9(2), 251-253 (2007-01-16)
A strong asymmetric amplification is observed in the addition of diethylzinc on aromatic aldehydes in the presence of the bistriflamide of trans-1,2-diaminocyclohexane 3a. The asymmetric amplification originates from the insolubility of the catalyst precursor 3a of low enantiomeric excess (ee)
Weimin Lin et al.
The Journal of organic chemistry, 74(2), 645-651 (2008-12-06)
The application of a zinc carbenoid-mediated chain-extension reaction to a functionalized peptide isostere is reported. The cleavage site of human CVM protease was utilized as a target for testing the synthetic methodology. The utility of this chain-extension reaction is demonstrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service