Skip to Content
Merck
All Photos(1)

Key Documents

255165

Sigma-Aldrich

Tributylmethylammonium chloride solution

75 wt. % in H2O

Synonym(s):

Methyltributylammonium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3N[CH3(CH2)3]3Cl
CAS Number:
Molecular Weight:
235.84
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

7.9 mmHg ( 25 °C)

concentration

75 wt. % in H2O

refractive index

n20/D 1.4682

bp

152 °C

density

0.964 g/mL at 25 °C

SMILES string

[Cl-].CCCC[N+](C)(CCCC)CCCC

InChI

1S/C13H30N.ClH/c1-5-8-11-14(4,12-9-6-2)13-10-7-3;/h5-13H2,1-4H3;1H/q+1;/p-1

InChI key

IPILPUZVTYHGIL-UHFFFAOYSA-M

Related Categories

General description

Tributylmethylammonium chloride (TBMAC) is a quaternary ammonium salt commonly used as a phase-transfer catalyst in various organic synthesis reactions. It can also be used in the preparation of ionic liquids.

Application

Tributylmethylammonium chloride solution is used as a phase transfer catalyst in the synthesis of ɛ-caprolactone by Baeyer-Villiger oxidation of cyclohexanone in the presence of KHSO5 as an oxidizing agent.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New and efficient technique for the synthesis of ?-caprolactone using KHSO5 as an oxidising agent in the presence of a phase transfer catalyst
Baj S, et al.
Applied Catalysis A: General, 395(1-2), 49-52 (2011)
Enantioselective biocatalytic reduction of 2 H-1, 4-benzoxazines using imine reductases
Zumbra?gel N, et al.
The Journal of Organic Chemistry, 84(3), 1440-1447 (2018)
J W Smit et al.
The Journal of pharmacology and experimental therapeutics, 286(1), 321-327 (1998-07-10)
We recently showed that absence of mdr1-type P-glycoprotein (P-gp) in mice resulted in profoundly reduced hepatic and intestinal clearance of type 1 and type 2 cationic drugs compared with that in wild-type mice. These data strongly support the concept that
Soon-Sun Hong et al.
Archives of pharmacal research, 29(4), 318-322 (2006-05-10)
Many quaternary ammonium salts are incompletely absorbed after their oral administration and may also be actively secreted into the intestine. However, the underlying mechanism(s) that control the transport of these cations across the intestinal epithelium is not well understood. In
Paula Ossowicz et al.
Molecules (Basel, Switzerland), 24(18) (2019-09-11)
Ionic liquids based on different l-amino acids (glycine, l-valine, l-leucine, l-isoleucine, l-histidine, l-methionine, l-tyrosine, l-tryptophan, l-arginine, and l-threonine) and different cations (tetrabutylammonium (TBA), tributylmethylammonium (tBMA), didecyldimethylammonium (DDA), (2-hydroxyethyl)trimethylammonium (choline) (Chol), alkyl(C12-C14) dimethylbenzylammonium (benzalkonium) (BA), dodecyltrimethylammonium (DDTMA), hexadecyltrimethylammonium (HDTMA), octadecyltrimethylammonium (ODTMA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service