254517
Trimethyl orthovalerate
97%
Synonym(s):
1,1,1-Trimethoxypentane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH2CH2CH2C(OCH3)3
CAS Number:
Molecular Weight:
162.23
Beilstein:
1739200
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.410 (lit.)
bp
164-166 °C (lit.)
density
0.941 g/mL at 25 °C (lit.)
functional group
ether
SMILES string
CCCCC(OC)(OC)OC
InChI
1S/C8H18O3/c1-5-6-7-8(9-2,10-3)11-4/h5-7H2,1-4H3
InChI key
XUXVVQKJULMMKX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Kinetics of unimolecular gas-phase elimination of trimethyl orthovalerate has been investigated.
Application
Trimethyl orthovalerate has been used in the preparation of:
- various 9-O-acyl derivatives of N-acetyl- and N-glycoloyl-neuraminic acid
- 2-methyl, 2-ethyl, 2-propyl and 2-butyl-3-benzimidazolyl-4(3H)-quinazolinones
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
107.6 °F - closed cup
Flash Point(C)
42 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Edgar Marquez et al.
The journal of physical chemistry. A, 114(12), 4203-4209 (2010-03-10)
The rates of gas-phase elimination of trimethyl orthovalerate and trimethyl orthochloroacetate have been determined in a static system, and the reaction Pyrex vessels have been deactivated with the product of decomposition of allyl bromide. The reactions are unimolecular and follow
H Ogura et al.
Carbohydrate research, 167, 77-86 (1987-09-15)
Various 9-O-acyl derivatives of N-acetyl- and N-glycoloyl-neuraminic acid, and O-(5-acetamido-3,5-dideoxy-D-glycero-alpha- and beta-D-galacto-2-nonulopyranosylonic acid)-(2----6)-O-beta-D-galactopyranosyl-(1----4)-D-glucopyranose were regioselectively synthesized by use of ortho esters. In addition, 5-acetamido-4-O-acetyl-D-glycero-D-galacto-2-nonulopyranosonic acid was prepared starting from the benzyl and methyl esters of N-acetylneuraminic acid.
A facile synthesis of new 3-(2-benzimidazolyl)-2-alkyl-4-(3H)-quinazolinones under microwave irradiation.
Hazarkhani H and Karimi B.
Tetrahedron, 59(26), 4757-4760 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service