Skip to Content
Merck
All Photos(1)

Key Documents

24880

Sigma-Aldrich

5-Chloro-7-iodo-8-quinolinol

≥95.0% (HPLC)

Synonym(s):

5-Chloro-8-hydroxy-7-iodoquinoline, Clioquinol, Iodochlorhydroxyquin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5ClINO
CAS Number:
Molecular Weight:
305.50
Beilstein:
153637
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (HPLC)

form

powder

solubility

dioxane: soluble 0.5 g/10 mL, clear, faintly yellow to yellow

functional group

chloro
iodo

SMILES string

Oc1c(I)cc(Cl)c2cccnc12

InChI

1S/C9H5ClINO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H

InChI key

QCDFBFJGMNKBDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Chloro-7-iodo-8-quinolinol is an antibiotic with metal-binding properties and exhibits anticancer activity in vitro and in vivo. 5-Chloro-7-iodo-8-quinolinol, its glucuronide and sulfate in serum, urine and milk has been quantitated by gas chromatography.

Application

5-Chloro-7-iodo-8-quinolinol has been used in analysis of intracellular free zinc ions with the FluoZin-3 probes using fluorescence microscopy.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A gas chromatographic determination method of 5-chloro-7-iodo-8-quinolinol and its conjugates in biological fluids.
C Chen et al.
Chemical & pharmaceutical bulletin, 24(1), 97-101 (1976-01-01)
Alessio Innocenti et al.
Bioorganic & medicinal chemistry letters, 18(5), 1583-1587 (2008-02-05)
The inhibition of the metalloenzyme carbonic anhydrase (CA, EC 4.2.1.1) with three phenols was investigated. Phenol was an effective CA I-IV, IX, XII and XIV inhibitor (K(I)s of 2.7-11.5 microM) and a less effective one against the other isoforms, CA
Silvio R Bareggi et al.
CNS neuroscience & therapeutics, 18(1), 41-46 (2011-01-05)
Clioquinol was produced as a topical antiseptic and marketed as an oral intestinal amebicide in 1934, being used to treat a wide range of intestinal diseases. In the early 1970s, it was withdrawn from the market as an oral agent
Jie Geng et al.
Advanced healthcare materials, 1(3), 332-336 (2012-11-28)
Metal ions play important roles in amyloid aggregation and neurotoxicity. Metal-ion chelation therapy has been used in clinical trials for Alzheimer's disease (AD) treatment. However, clinical trial studies have shown that long-term use of metal chelator can cause adverse side
Lin W Hung et al.
Future medicinal chemistry, 4(8), 955-969 (2012-06-02)
In 1906, Alois Alzheimer first characterized the disease that bears his name. Despite intensive research, which has led to a better understanding of the pathology, there is no effective treatment for this disease. Of the drugs approved by the US

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service