244740
Methyl 4-formylbenzoate
99%
Synonym(s):
Methyl benzaldehyde-4-carboxylate, Methyl terephthalaldehydate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
HCOC6H4CO2CH3
CAS Number:
Molecular Weight:
164.16
Beilstein:
775895
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
99%
bp
265 °C (lit.)
mp
59-63 °C (lit.)
functional group
aldehyde
ester
SMILES string
[H]C(=O)c1ccc(cc1)C(=O)OC
InChI
1S/C9H8O3/c1-12-9(11)8-4-2-7(6-10)3-5-8/h2-6H,1H3
InChI key
FEIOASZZURHTHB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
262.4 °F
Flash Point(C)
128 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xinping Liu et al.
ACS nano, 13(5), 5222-5230 (2019-04-20)
Metal-organic frameworks (MOFs)-based peroxidase mimics have been seldom applied in the biomedical field, especially in vivo. One of the main reasons is their optimum reactions occur in strong acidic environments with a pH of 3-4, severely limiting their applications in
Ole Winkelmann et al.
Organic & biomolecular chemistry, 7(3), 553-556 (2009-01-22)
Two bimacrocyclic imidazolinium salts of different size, precursors to respective NHCs (N-heterocyclic carbenes), were tested as precatalysts in the reaction of aromatic aldehydes or ketones with enals. The expected lactones were produced in most cases, but in the reaction of
Yijun Deng et al.
Journal of medicinal chemistry, 51(16), 5052-5063 (2008-08-06)
6-Substituted classical pyrrolo[2,3-d]pyrimidine antifolates with a three- to six-carbon bridge between the heterocycle and the benzoyl-L-glutamate (compounds 2-5, respectively) were synthesized starting from methyl 4-formylbenzoate and a Wittig reaction with the appropriate triphenylphosphonium bromide, followed by reduction and conversion to
Naoya Kishikawa et al.
Phytochemical analysis : PCA, 29(1), 112-117 (2017-09-13)
Tanshinones are a major class of bioactive ingredients in the traditional herbal medicines, Danshen (Salvia miltiorrhiza). A sensitive and reliable determination method for tanshinones is useful to ensure the quality of Danshen. To develop a sensitive and selective analytical method
Mahmoud Elcheikh Mahmoud et al.
Journal of the American Chemical Society, 141(17), 7115-7121 (2019-04-12)
In this study, we report the successful incorporation of the photoactive bis(4'-(4-carboxyphenyl)-terpyridine)ruthenium(II) (Ru(cptpy)2) strut into a robust metal-organic framework (MOF), AUBM-4. The single crystal X-ray analysis revealed the formation of a new one-dimensional structure of Ru(cptpy)2 complexes linked together by Zr
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service