Skip to Content
Merck
All Photos(2)

Documents

244325

Sigma-Aldrich

2-Iodobutane

≥98%, contains copper as stabilizer

Synonym(s):

sec-Butyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CHICH3
CAS Number:
Molecular Weight:
184.02
Beilstein:
1718777
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.499 (lit.)

bp

119-120 °C (lit.)

mp

−104 °C (lit.)

solubility

alcohol: soluble(lit.)
diethyl ether: soluble(lit.)
water: insoluble(lit.)

density

1.598 g/mL at 25 °C (lit.)

SMILES string

CCC(C)I

InChI

1S/C4H9I/c1-3-4(2)5/h4H,3H2,1-2H3

InChI key

IQRUSQUYPCHEKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Iodobutane was degraded by reductive dehalogenation with nickel-aluminum alloy in potassium hydroxide solution. The release rate of 2-iodobutane, a volatile organoiodide, was studied.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kajari Chatterjee et al.
ACS omega, 5(27), 16681-16689 (2020-07-21)
Our society is critically dependent on lithium-ion batteries (LIBs) as a power source for portable electronic gadgets. One of the major problems with these batteries is the degradation of the materials inside them. In addition to the reduced cell life
Frank Keppler et al.
Chemosphere, 52(2), 477-483 (2003-05-10)
Volatile iodinated organic compounds play an important role in the tropospheric photochemical system, but the current knowledge of the known sources and sinks of these alkyl iodides is still incomplete. This paper describes a new source of alkyl iodides from
Kajari Chatterjee et al.
Scientific reports, 10(1), 9606-9606 (2020-06-17)
A novel dicationic room temperature ionic liquid, 1,1'-(5,14-dioxo-4,6,13,15-tetraazaoctadecane-1,18-diyl) bis(3-(sec-butyl)-1H-imidazol-3-ium) bis((trifluoromethyl)-sulfonyl) imide has been synthesized and fully characterized. Its thermal and electrochemical analyses along with transport properties have been studied. We propose it as a potential nominal additive to the commonly
G Lunn et al.
American Industrial Hygiene Association journal, 52(6), 252-257 (1991-06-01)
Two techniques were investigated for degrading a number of halogenated compounds of commercial and research importance. Reductive dehalogenation with nickel-aluminum alloy in potassium hydroxide solution was used to degrade iodomethane, chloroacetic acid, trichloroacetic acid, 2-chloroethanol, 2-bromoethanol, 2-chloroethylamine, 2-bromoethylamine, 1-bromobutane, 1-iodobutane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service