Skip to Content
Merck
All Photos(2)

Documents

237817

Sigma-Aldrich

D-(−)-Pantolactone

99%

Synonym(s):

(R)-(−)-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, (R)-(−)-β,β-Dimethyl-α-hydroxy-γ-butyrolactone, (R)-(−)-Pantolactone, Pantoic acid γ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
Beilstein:
80957
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

optical activity

[α]25/D −49.8°, c = 2 in H2O

bp

120-122 °C/15 mmHg (lit.)

mp

91 °C (lit.)

SMILES string

CC1(C)COC(=O)[C@@H]1O

InChI

1S/C6H10O3/c1-6(2)3-9-5(8)4(6)7/h4,7H,3H2,1-2H3/t4-/m0/s1

InChI key

SERHXTVXHNVDKA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(-)-Pantolactone is a chiral auxiliary used in many asymmetric synthesis reactions.
It can be used as a chiral starting material to synthesize:
  • An insect sex pheromone named 1S,2S,3R-1-acetoxymethyl-2,3,4,4-tetrameth-ylcyclopentane.
  • (-)-Enantiomer of (R)-8-hydroxy-4,7,7-trimethyl-7,8-dihydrocyclopenta[e]isoindole-1,3(2H,6H)-dione, a norsesquiterpene alkaloid.
  • A bicyclic diterpene named isofregenedadiol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of isofregenedadiol
Kurhade SE, et al.
Organic Letters, 13(14), 3690-3693 (2011)
Enantiospecific synthesis of sex pheromone of the obscure mealybug from pantolactone via tandem conjugate addition/cyclization
Hajare AK, et al.
Tetrahedron Letters, 51(40), 5291-5293 (2010)
Asymmetric diels alder reactions and michael type additions with 6 (R)-3′(R)-Pantolactone-substituted-2H-pyran-3 (6H)-one.
Knol J, et al.
Tetrahedron Letters, 32(50), 7465-7468 (1991)
Total synthesis of an anticancer norsesquiterpene alkaloid isolated from the fungus Flammulina velutipes
Kashinath K, et al.
Organic & Biomolecular Chemistry, 12(24), 4098-4103 (2014)
Diastereoface-discriminative metal coordination in asymmetric synthesis: d-pantolactone as practical chiral auxiliary for Lewis acid catalyzed Diels-Alder reactions
Poll T, et al.
Tetrahedron Letters, 26(26), 3095-3098 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service