Skip to Content
Merck
All Photos(1)

Key Documents

236691

Sigma-Aldrich

(R)-(−)-2-Butanol

99%

Synonym(s):

(R)-(−)-sec-Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(OH)CH3
CAS Number:
Molecular Weight:
74.12
Beilstein:
1718764
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.6 (vs air)

vapor pressure

12.5 mmHg ( 20 °C)

Assay

99%

form

liquid

optical activity

[α]20/D −13°, neat

autoignition temp.

761 °F

expl. lim.

9.8 %

refractive index

n20/D 1.397 (lit.)

bp

97-100 °C (lit.)

density

0.807 g/mL at 20 °C (lit.)

functional group

hydroxyl

SMILES string

CC[C@@H](C)O

InChI

1S/C4H10O/c1-3-4(2)5/h4-5H,3H2,1-2H3/t4-/m1/s1

InChI key

BTANRVKWQNVYAZ-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(−)-2-Butanol can be used as a reagent in the synthesis of:
  • 1-[(S)-2-butyl]-3,5 diphenyl-1H-1,2,4 triazole from 3,5 diphenyl-1H-1,2,4 triazole by Mitsunobu reaction.
  • 2-butyl tosylate building block by reacting with toluenesulfonyl chloride.

(R)-(-)-2-Butanol has been used as a freeze drying medium during the long term cryopreservation of smooth muscle cells samples in a freezing container.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K Kulyk et al.
Journal of the American Society for Mass Spectrometry, 28(12), 2686-2691 (2017-09-25)
Here we report on the gas-phase interactions between protonated enantiopure amino acids (L- and D-enantiomers of Met, Phe, and Trp) and chiral target gases [(R)- and (S)-2-butanol, and (S)-1-phenylethanol] in 0.1-10.0 eV low-energy collisions. Two major processes are seen to
Irene O C M Vroegrijk et al.
Obesity (Silver Spring, Md.), 21(10), 2037-2045 (2013-03-21)
The scavenger receptor CD36 facilitates the cellular uptake of long-chain fatty acids. As CD36-deficiency attenuates the development of high fat diet (HFD)-induced obesity, the role of CD36-deficiency in preadipocyte recruitment and adipocyte function was set out to characterize. Fat cell
Stereochemistry of trifluoroacetolysis of optically active 2-Butyl Tosylate. A test for hydrogen bridging
Allen AD, et al.
The Journal of Organic Chemistry, 48(24), 4527-4530 (1983)
S Kooijman et al.
Journal of thrombosis and haemostasis : JTH, 13(1), 126-135 (2014-10-28)
The autonomic nervous system attenuates inflammation through activation of the α7 nicotinic acetylcholine receptor (α7nAChR), a pathway termed the cholinergic anti-inflammatory reflex. Interestingly, α7nAChR is expressed on immune cells and platelets, both of which play a crucial role in the
Richard G Ijzerman et al.
Pediatric research, 52(6), 868-872 (2002-11-20)
Low birth weight may be associated with high levels of cholesterol in later life through genetic factors that affect both birth weight and cholesterol metabolism. Alterations in cholesterol synthesis and absorption may play an important role in this association. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service