Skip to Content
Merck
All Photos(1)

Key Documents

236535

Sigma-Aldrich

Fluorosulfonic acid

purified by triple-distillation

Synonym(s):

Fluosulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FSO3H
CAS Number:
Molecular Weight:
100.07
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

vapor density

3.5 (vs air)

vapor pressure

2.5 mmHg ( 25 °C)

purified by

triple-distillation

bp

165.5 °C (lit.)

mp

−87.3 °C (lit.)

density

1.726 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OS(F)(=O)=O

InChI

1S/FHO3S/c1-5(2,3)4/h(H,2,3,4)

InChI key

UQSQSQZYBQSBJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Polymerization of epoxidized triglycerides with fluorosulfonic acid: The article investigates the use of fluorosulfonic acid for the cationic polymerization of epoxidized triglycerides, exploring its potential in polymer chemistry (Biswas et al., 2016).

Packaging

Packaged in PFA/FEP bottles

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Renosto et al.
The Journal of biological chemistry, 262(34), 16279-16288 (1987-12-05)
ATP sulfurylase from Penicillium chrysogenum is a noncooperative homooligomer containing three free sulfhydryl groups per subunit. Under nondenaturing conditions, one SH group per subunit was modified by 5,5'-dithiobis-(2-nitrobenzoate), or N-ethylmaleimide. Modification had only a small effect on kcat, but markedly
Leslie Dos Santos et al.
Membranes, 10(10) (2020-09-27)
Electrospinning was employed to fabricate composite membranes containing perfluorosulfonic acid (PFSA) ionomer, poly(vinylidene fluoride) (PVDF) reinforcement and a sulfonated silica network, where the latter was incorporated either in the PFSA matrix or in the PVDF fibers. The best membrane, in
Roger L Snowden
Chemistry & biodiversity, 5(6), 958-969 (2008-07-12)
Using a novel, acid-mediated cyclization methodology, a direct access to Cetalox ((+/-)-1; a commercially important ambergris-type odorant) and various structurally related didehydro (i.e., 19, 26, and 30) and tetradehydro (i.e., 28 and 37/38) analogues is described. Treatment of either (E,E)-14
Xiaolu Li et al.
Analytica chimica acta, 1122, 31-38 (2020-06-07)
An upright GO (UGO) modified screen-printed electrode was prepared with the help of the external magnetic field for improving its electrochemical performance. The ratio of GO: Nafion and the magnetic field intensity on the properties of UGO were examined by
Baojian Zhang et al.
Mikrochimica acta, 185(8), 385-385 (2018-07-26)
A nickel nanoparticle/nafion-graphene oxide (NiNP/Nf-GO) modified screen-printed electrode (SPE) was developed for rapid and environmentally friendly electrochemical determination of chemical oxygen demand (COD). The morphology and the electrochemical performance of the SPEs with different surface modifications were investigated by scanning electron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service