Skip to Content
Merck
All Photos(2)

Documents

235725

Sigma-Aldrich

Trichloro(octyl)silane

97%

Synonym(s):

Octyltrichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7SiCl3
CAS Number:
Molecular Weight:
247.67
Beilstein:
1744230
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

Assay

97%

form

liquid

refractive index

n20/D 1.447 (lit.)

bp

233 °C/731 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC[Si](Cl)(Cl)Cl

InChI

1S/C8H17Cl3Si/c1-2-3-4-5-6-7-8-12(9,10)11/h2-8H2,1H3

InChI key

RCHUVCPBWWSUMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloro(octyl)silane (OTS) is a linear alkylsilane which is used as a modifying agent which creates a low cost hydrophobic surface with high thermo-mechanical stability. OTS contains hydrolysable groups which form a covalent bond with the hydroxyl groups present on the surface.

Application

OTS functionalized magnesium oxide (MnO2)/aluminium oxide (Al2O3) surface can be used as a catalyst in the decomposition of ozone (O3) and nitrogen dioxide (NO2). OTS can also be used for the surface modification of silicon oxide (SiO2), used in the fabrication of pentacene organic field effect transistors (OFETs). OTS functionalized wafers can be coated on enhanced green fluorescent protein (GFP) and anchor peptides based films to determine the thickness of the films.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn F A Clancy et al.
Biosensors & bioelectronics, 130, 397-407 (2018-09-27)
The uniformity of the protein patterns, their shape, and the contrast between the fluorescence signal of the pattern and the background, critically modulate the quantitative accuracy of the microarray-derived data. While significant research focused of the identification of the factors
Shuichi Nagamatsu et al.
Materials (Basel, Switzerland), 12(21) (2019-11-07)
A novel film preparation method utilizing polymer suspension, entitled adsorbing deposition in suspensions (ADS), has been proposed. The poly(3-hexylthiophene) (P3HT) toluene solution forms P3HT nanofibrils dispersed suspension by aging. P3HT nanofibrils are highly crystallized with sharp vibronic absorption spectra. By
Kechun Zhang et al.
Journal of the American Chemical Society, 127(29), 10136-10137 (2005-07-21)
An artificial polypeptide scaffold composed of surface anchor and protein capture domains was designed and expressed in vivo. By using a mutant E. coli phenylalanyl-tRNA synthetase, the photoreactive amino acid para-azidophenylalanine was incorporated into the surface anchor domain. Octyltrichlorosilane-treated surfaces
Anchor peptides: A green and versatile method for polypropylene functionalization.
Rubsam K, et al.
Polymer, 116(1), 124-132 (2017)
Performance comparison of pentacene organic field-effect transistors with SiO2 modified with octyltrichlorosilane or octadecyltrichlorosilane.
Tiwari SP.
Organic Electronics, 13(1), 18-22 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service