Skip to Content
Merck
All Photos(2)

Documents

235199

Sigma-Aldrich

7-Methoxycoumarin-4-acetic acid

97%

Synonym(s):

7-Methoxy-2-oxo-2H-1-benzopyran-4-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O5
CAS Number:
Molecular Weight:
234.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

193 °C (dec.) (lit.)

solubility

DMF: soluble 50 mg/mL, clear, colorless to yellow

SMILES string

COc1ccc2C(CC(O)=O)=CC(=O)Oc2c1

InChI

1S/C12H10O5/c1-16-8-2-3-9-7(4-11(13)14)5-12(15)17-10(9)6-8/h2-3,5-6H,4H2,1H3,(H,13,14)

InChI key

ZEKAXIFHLIITGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

7-Methoxycoumarin-4-acetic acid has been used:
  • as fluorescent probe in preparation of two novel LysB29 selectively labelled fluorescent derivatives of human insulin
  • in the preparation of cell-penetrating peptide transportan 10 (tp10)
  • as fluorescent label for peptides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianshu Xiao et al.
Nature structural & molecular biology, 28(2), 202-209 (2021-01-13)
Effective intervention strategies are urgently needed to control the COVID-19 pandemic. Human angiotensin-converting enzyme 2 (ACE2) is a membrane-bound carboxypeptidase that forms a dimer and serves as the cellular receptor for severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2). ACE2 is
Alice Ciencialová et al.
Journal of peptide science : an official publication of the European Peptide Society, 10(7), 470-478 (2004-08-10)
The preparation and characterization of two novel LysB29 selectively labelled fluorescent derivatives of human insulin are described. Two probes were chosen: 4-chloro-7-nitrobenz-2-oxa-1,3-diazole (NBD) and 7-methoxycoumarin-4-acetic acid (MCA), which have a relatively small, compact structure and are able to react with
Stereospecific synthesis ofl-2-amino-3-(7-methoxy-4-coumaryl) propionic acid, an alternative to tryptophan in quenched fluorescent substrates for peptidases.
Knight CG.
Lett. Pept. Sci., 5(1), 1-4 (1998)
Lindsay E Yandek et al.
Biophysical journal, 92(7), 2434-2444 (2007-01-16)
The mechanism of the interaction between the cell-penetrating peptide transportan 10 (tp10) and phospholipid membranes was investigated. Tp10 induces graded release of the contents of phospholipid vesicles. The kinetics of peptide association with vesicles and peptide-induced dye efflux from the
Hisatsugu Koshimizu et al.
Molecular brain, 2, 27-27 (2009-08-14)
Proneurotrophins and mature neurotrophins elicit opposite effects via the p75 neurotrophin receptor (p75(NTR)) and Trk tyrosine kinase receptors, respectively; however the molecular roles of proneurotrophins in the CNS are not fully understood. Based on two rare single nucleotide polymorphisms (SNPs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service