Skip to Content
Merck
All Photos(1)

Key Documents

234273

Sigma-Aldrich

trans,trans,trans-1,5,9-Cyclododecatriene

99%

Synonym(s):

(1E,5E,9E)-1,5,9-Cyclododecatriene, (1E,5E,9E)-Cyclododeca-1,5,9-triene, (E,E,E)-1,5,9-Cyclododecatriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18
CAS Number:
Molecular Weight:
162.27
Beilstein:
969880
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

237-238 °C (lit.)

mp

33-35 °C (lit.)

SMILES string

C1C\C=C\CC\C=C\CC\C=C\1

InChI

1S/C12H18/c1-2-4-6-8-10-12-11-9-7-5-3-1/h1-2,7-10H,3-6,11-12H2/b2-1+,9-7+,10-8+

InChI key

ZOLLIQAKMYWTBR-FFWAUJBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans,trans,trans-1,5,9-Cyclododecatriene (ttt-cdt) acts as ligand and forms rare copper complexes. Photorearrangements of CuCl complexes of ttt-cdt has been investigated.

Application

trans,trans,trans-1,5,9-Cyclododecatriene has been used in the construction of building block for the synthesis of potent antitumor annonaceous acetogenin (+)-parviflorin.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mauro Fianchini et al.
Dalton transactions (Cambridge, England : 2003), (12)(12), 2085-2087 (2009-03-11)
Two rare copper complexes, a tri-alkene adduct [Cu(ttt-cdt)(FSbF(5))] and a non-classical carbon monoxide complex [Cu(ttt-cdt)(CO)][SbF(6)] with a high CO stretching frequency ([small nu, Greek, macron](CO) = 2160 cm(-1)), have been isolated using trans,trans,trans-1,5,9-cyclododecatriene (ttt-cdt) ligand.
Highly efficient synthesis of the potent antitumor annonaceous acetogenin (+)-parviflorin.
Hoye TR and Ye Z.
Journal of the American Chemical Society, 118(7), 1801-1802 (1996)
Photorearrangements of CuCl complexes of 1, 5, 9-cyclododecatrienes: wavelength dependence.
Chow YL, et al.
Organometallics, 6(5), 1126-1129 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service