Skip to Content
Merck
All Photos(1)

Key Documents

227730

Sigma-Aldrich

Silver hexafluoroantimonate(V)

98%

Synonym(s):

Silver hexafluoroantimony, Silver hexafluorostiboranuide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgSbF6
CAS Number:
Molecular Weight:
343.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

Quality Level

Assay

98%

form

powder

reaction suitability

reagent type: catalyst
core: silver

SMILES string

[Ag+].F[Sb-](F)(F)(F)(F)F

InChI

1S/Ag.6FH.Sb/h;6*1H;/q+1;;;;;;;+5/p-6

InChI key

GSXFODUDOAXUBF-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Generates stable cation radical salts and is used as an acidic catalyst in epoxide opening reactions and sulfenylation reactions. Used to prepare silver 3,3′-dicyanodiphenylacetylene coordination networks for study of the effect of ligands on network structure toward the goal of engineering novel materials.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry Letters (Jpn), 1901-1901 (1992)
Development of a Nazarov cyclization/Wagner-Meerwein rearrangement sequence for the stereoselective synthesis of spirocycles.
Jie Huang et al.
Journal of the American Chemical Society, 129(26), 8060-8061 (2007-06-09)
The Journal of Organic Chemistry, 57, 6178-6178 (1992)
Chemistry Letters (Jpn), 1-1 (1993)
Keith A. Hirsch et al.
Inorganic chemistry, 36(14), 2960-2968 (1997-07-02)
Coordination networks of 3,3'-dicyanodiphenylacetylene (3,3'-DCPA, 1) with silver(I) salts characterized by single-crystal X-ray analysis are described. Network topology is found to depend on both the counterion and solvent employed during crystallization. The conformation adopted by the ligand varies between planar

Articles

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service