Skip to Content
Merck
All Photos(3)

Key Documents

221171

Sigma-Aldrich

Pinacol

98%

Synonym(s):

2,3-Dimethyl-2,3-butanediol, Tetramethylethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(CH3)2C(CH3)2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1340501
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

solid

bp

171-172 °C/739 mmHg (lit.)

mp

40-43 °C (lit.)

solubility

alcohol: freely soluble
diethyl ether: freely soluble
hot water: freely soluble

functional group

hydroxyl

SMILES string

CC(C)(O)C(C)(C)O

InChI

1S/C6H14O2/c1-5(2,7)6(3,4)8/h7-8H,1-4H3

InChI key

IVDFJHOHABJVEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The pinacol coupling reaction with the inorganic electride [Ca2N](+)·e(−) as an electron donor in organic solvents was studied[1].

Application

Pinacol is a 1,2-diol that can be used:
  • As a ligand to prepare uranyl complexes by reacting with uranyl nitrate and acetate hydrates.[2]
  • To prepare pinacolone and 2,3-dimethyl-1,3-butadiene by typical pinacol rearrangement reaction.[3][4]
  • As a reactant to synthesize 4-(4,4,5,5-tetramethyl-1,3,2-bioxaborolan-2-yl)benzoic acid, which is used to prepare derivatives of 4-acetoxy-3-phenylbenzaldehyde.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

New extraction procedure for protonated polyoxometalates prepared in aqueous-organic solution and characterisation of their catalytic ability
Ueda T, et al.
Applied Catalysis A: General, 485(1-2), 181-187 (2014)
Synthesis of polymer-bound 4-acetoxy-3-phenylbenzaldehyde derivatives: applications in solid-phase organic synthesis
Kumar A, et al.
The Journal of Organic Chemistry, 71(20), 7915-7918 (2006)
Uranyl complexes with 1, 2-diols and tetrahydrofurfuryl alcohols
Villiers C, et al.
Polyhedron, 46(1), 133-138 (2012)
Fe-substituted molecular sieves as catalysts in liquid phase pinacol rearrangement
Hsien M, et al.
J. Mol. Catal. A: Chem., 181(1-2), 189-200 (2002)
Ye Ji Kim et al.
Chemical communications (Cambridge, England), 50(37), 4791-4794 (2014-04-02)
The scalable pinacol coupling reaction is realized utilizing the inorganic electride [Ca2N](+)·e(-) as an electron donor in organic solvents. The bond cleavages of the [Ca2N](+) layers by methanol play a vital role in transferring anionic electrons to electrophilic aldehydes, accompanying

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service