Skip to Content
Merck
All Photos(3)

Key Documents

218928

Sigma-Aldrich

Sodium dichloroisocyanurate

96%

Synonym(s):

NaDCC, 3,5-Dichloro-2-hydroxy-4,6-s-triazinedione sodium salt, Dichloro-s-triazinetrione sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3Cl2N3NaO3
CAS Number:
Molecular Weight:
219.95
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

(Granules or Granular Powder)

reaction suitability

reagent type: oxidant

SMILES string

[Na]N1C(=O)N(Cl)C(=O)N(Cl)C1=O

InChI

1S/C3HCl2N3O3.Na/c4-7-1(9)6-2(10)8(5)3(7)11;/h(H,6,9,10);/q;+1/p-1

InChI key

MSFGZHUJTJBYFA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium dichloroisocyanurate (NaDCC) is also used as a precursor to produce hypochlorous acid, a known oxidizing agent used in organic synthesis.

Application

-Reactant or reagent for:

  • N-monochlorination and dehydrochlorination of amino esters
  • Chlorination to detect ammonium via formation of colored zebra-bands in a deteting tube
  • Synthesis of antimicrobial polyurethane additives to control biofilm
  • Preparation of bromochloroisocyanurate for electrophilic cobromination of alkenes and bromination of arenes
NaDCC can be used as:        
  • An oxidizing reagent for the preparation of tetrazene(N–N=N–N)-linked bi(1,2,4-triazole) from 4-amino-1,2,4-triazole by oxidizing N-NH2 functional group.        
  • A chlorinating agent for the N-monochlorination of α-amino esters and α-amino-β-diesters.

It can also be used to prepare NaDCC immobilized magnetic nano-Fe3O4 material, which is used as an excellent disinfectant as well as a wastewater treatment agent.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of new tetrazene (N-NN-N)-linked bi (1, 2, 4-triazole)
Li Sheng H, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 18(10), 1176-1178 (2007)
Anita Rácz et al.
Zebrafish, 16(1), 77-86 (2018-10-26)
In 2017, the zebrafish unit at University of Glasgow experienced a detrimental outbreak of pathogenic bacterium, Mycobacterium haemophilum. The presence of other bacterial species was also confirmed by bacteriology growth in the same unit. The affected individuals composed of a
Steffen Werner et al.
Environmental pollution (Barking, Essex : 1987), 233, 561-568 (2017-11-06)
In many water-scarce countries, waste water is used for irrigation which poses a health risk to farmers and consumers. At the same time, it delivers nutrients to the farming systems. In this study, we tested the hypotheses that biochar can
A highly efficient method for the ?, ?-dehydrogenation of ? amino esters and ?-amino- ? -diesters
Pallavicini, M, et al.
Tetrahedron Letters, 51(42), 5540-5542 (2010)
Lang Gong et al.
BMC veterinary research, 16(1), 358-358 (2020-09-27)
Pseudorabies (PR) is latent and can persist in infected sows for a long time, and thus, convalescent sows can carry the virus throughout life, causing severe economic losses to farmers and posing a tremendous challenge to PR prevention and control.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service