Skip to Content
Merck
All Photos(3)

Key Documents

210633

Sigma-Aldrich

Acid Red 1

Dye content 60 %

Synonym(s):

Amido Naphthol Red G, Azophloxine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13N3Na2O8S2
CAS Number:
Molecular Weight:
509.42
Colour Index Number:
18050
Beilstein:
4121767
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 60%

λmax

506 nm
532 nm (2nd)

ε (extinction coefficient)

≥17000 at 529-535 nm in water at 0.02 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].CC(=O)Nc1cc(cc2cc(c(\N=N\c3ccccc3)c(O)c12)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C18H15N3O8S2.2Na/c1-10(22)19-14-9-13(30(24,25)26)7-11-8-15(31(27,28)29)17(18(23)16(11)14)21-20-12-5-3-2-4-6-12;;/h2-9,23H,1H3,(H,19,22)(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2/b21-20+;;

InChI key

WXLFIFHRGFOVCD-SERMZQFOSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Histochemical detection of myoepithelial cells in pleomorphic adenoma with azophloxine and amidoblack.
A G Triantafyllou
Journal of oral medicine, 41(3), 184-188 (1986-07-01)
C N Edwards et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 22(8), 593-597 (1984-08-01)
Urine and faecal extracts from rats given Brown FK or Red 2G orally (800 mg/kg body weight) were investigated for mutagenicity. Extracts were subjected to liquid fluctuation and plate incorporation assays with Salmonella typhimurium strains TA98 and TA100 in the
[Azofloxin--toluidine blue staining].
M Krutsay
Morphologiai es igazsagugyi orvosi szemle, 23(1), 39-40 (1983-01-01)
Qin Wei et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 22(2), 275-279 (2006-03-04)
A simple and sensitive method was conducted for the determination of trace amounts of proteins with benzeneazo-8-acetylamino-1-naphthol-3,6-disulfonic acid sodium salt (azophloxine, AP) using a Rayleigh light-scattering (RLS) technique. At pH 2.60 and in the presence of an emulsifier OP microemulsion
C N Edwards et al.
Mutation research, 117(1-2), 127-134 (1983-04-01)
The UK-permitted, monoazo food colour Red 2G (C.I. 18050) has previously been shown to induce transition mutations in bacteria when tested with metabolic activation. Whilst this result has now been confirmed, later commercial food-grade batches of the dye have also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service