Skip to Content
Merck
All Photos(1)

Key Documents

205850

Sigma-Aldrich

Ammonium tetrachloropalladate(II)

97%

Synonym(s):

Palladium(II)-ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2PdCl4
CAS Number:
Molecular Weight:
284.31
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
97%
form:
solid

Quality Level

Assay

97%

form

solid

density

2.17 g/mL at 25 °C (lit.)

SMILES string

[H][N+]([H])([H])[H].[H][N+]([H])([H])[H].Cl[Pd--](Cl)(Cl)Cl

InChI

1S/4ClH.2H3N.Pd/h4*1H;2*1H3;/q;;;;;;+2/p-2

InChI key

IRKVTUVFHGUMMN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Reacts with creatinine (cat. no. 859702) to form various Pd-creatinine complexes with applications in semiconductors, catalysis, mixed oxidation state studies, and electron conductivity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

You Yu et al.
Small (Weinheim an der Bergstrasse, Germany), 11(28), 3444-3449 (2015-03-20)
Stretchable and transparent electrodes are fabricated by chemical deposition of metal thin films on natural veins of leaves at ambient conditions. These vein-based transparent electrodes show excellent electro-optical property (0.9 Ω sq(-1) at 83% T) even at 50% tensile strains
Joris Muris et al.
Contact dermatitis, 67(2), 94-100 (2012-04-03)
Exposure to palladium (Pd) may lead to clinical allergic reactions. With frequent nickel (Ni) exposure and cross-reactivity between Ni and Pd at the T cell recognition level, positive Pd reactions on patch testing are surprisingly uncommon. PdCl(2) is often used
L V Tat'ianenko et al.
Voprosy meditsinskoi khimii, 28(6), 126-131 (1982-11-01)
Mechanism of interaction between biologically active substances and membranes as well as and membrane-bound enzymes of two types: mitochondrial monoamine oxidase (MAO) from rat liver tissue and Ca2+, Mg+2-dependent ATPase from sarcoplasmic reticulum (SR) were studied. All the substances studied
B L Iverson et al.
Nucleic acids research, 15(19), 7823-7830 (1987-10-12)
Reaction of DNA with K2PdCl4 at pH 2.0 followed by a piperidine workup produces specific cleavage at adenine (A) residues. Product analysis revealed the K2PdCl4 reaction involves selective depurination at adenine, affording an excision reaction analogous to the other chemical
G Pneumatikakis et al.
Journal of inorganic biochemistry, 37(1), 17-28 (1989-09-01)
The interactions of potassium tetrachloropalladate(II) with the B6 vitamins pyridoxal, pyridoxine, and pyridoxamine in 1:1 molar ratio have been studied. From DMF solutions, the ionic trichloro (pyridoxal or pyridoxine) palladates(II) were isolated. Pyridoxamine, on the other hand, in aqueous solutions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service