Skip to Content
Merck
All Photos(4)

Key Documents

204358

Sigma-Aldrich

Sand, white quartz

≥99.995% trace metals basis

Synonym(s):

Quartz, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99.995% trace metals basis

form

powder

application(s)

battery manufacturing

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

May contain adsorbed H2O and CO2 which is removable by calcining at >900°C

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soania Mathur et al.
Journal of Parkinson's disease, 5(2), 263-268 (2015-02-28)
Despite an urgent need for new medications, clinical trials in Parkinson's have a relatively low rate of success. Although many reasons have been proposed for this, the opinions of patients and scientists, the two principal stakeholders, have not been widely
Keren B Turton et al.
PloS one, 10(5), e0127243-e0127243 (2015-05-20)
Signal transducer and activator of transcription 3 (STAT3) is a key mediator of leukocyte differentiation and proliferation. The 3' end of STAT3 transcripts is subject to two alternative splicing events. One results in either full-length STAT3α or in STAT3β, which
Yijun Yao et al.
Journal of hazardous materials, 298, 221-231 (2015-06-10)
In 2002, U.S. EPA proposed a general buffer zone of approximately 100 feet (30 m) laterally to determine which buildings to include in vapor intrusion (VI) investigations. However, this screening distance can be threatened by factors such as extensive surface
C Uhrenfeldt et al.
Optics express, 23(11), A525-A538 (2015-06-16)
Plasmonic resonances in metal nanoparticles are considered candidates for improved thin film Si photovoltaics. In periodic arrays the influence of collective modes can enhance the resonant properties of such arrays. We have investigated the use of periodic arrays of Al
Manuela Piazzi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(4), 1383-1394 (2015-01-01)
The phosphoinositide-dependent signal transduction pathway has been implicated in the control of a variety of biologic processes, such as the regulation of cellular metabolism and homeostasis, cell proliferation and differentiation, and apoptosis. One of the key players in the regulation

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service