Skip to Content
Merck
All Photos(3)

Key Documents

204307

Sigma-Aldrich

Selenium

pellets, <5 mm particle size, ≥99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Se
CAS Number:
Molecular Weight:
78.96
EC Number:
MDL number:
UNSPSC Code:
12141910
PubChem Substance ID:
NACRES:
NA.23

description

gray

Assay

≥99.999% trace metals basis

form

pellets

resistivity

1.2 μΩ-cm, 0°C

particle size

<5 mm

bp

684.9 °C (lit.)

mp

217 °C (lit.)

density

4.81 g/mL at 25 °C (lit.)

SMILES string

[Se]

InChI

1S/Se

InChI key

BUGBHKTXTAQXES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Selenium is a non-metallic p-type semiconductor and is commercially produced as a by-product in refining sulfide-bearing copper minerals. It has good photovoltaic and photoconductive properties and is extensively used in the electronics such as solar cells, light meters, and, photocells. It also finds application in the field of ceramics, pigments, metallurgy, agriculture, glasses, and lubricants.

Application

Selenium has been used as a starting material tosynthesize selenium nanoparticles.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 4 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Waleed Alhazzani et al.
Critical care medicine, 41(6), 1555-1564 (2013-04-17)
Patients with sepsis syndrome commonly have low serum selenium levels. Several randomized controlled trials have examined the efficacy of selenium supplementation on mortality in patients with sepsis. To determine the efficacy and safety of high-dose selenium supplementation compared to placebo
Kai-Oliver Feldmann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(27), 9697-9712 (2015-05-12)
Although a fairly large number of binary group 15/16 element cations have been reported, no example involving phosphorus in combination with a group 16 element has been synthesized and characterized to date. In this contribution is reported the synthesis and structural characterization
Claire M Weekley et al.
Nutrients, 5(5), 1734-1756 (2013-05-24)
Determining the speciation of selenium in vivo is crucial to understanding the biological activity of this essential element, which is a popular dietary supplement due to its anti-cancer properties. Hyphenated techniques that combine separation and detection methods are traditionally and
Daniel D Karp et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(33), 4179-4187 (2013-09-05)
Selenium has been reported to have chemopreventive benefits in lung cancer. We conducted a double-blind, placebo-controlled trial to evaluate the incidence of second primary tumors (SPTs) in patients with resected non-small-cell lung cancer (NSCLC) receiving selenium supplementation. Patients with completely
Matthew Loxham et al.
Toxicological sciences : an official journal of the Society of Toxicology, 145(1), 98-107 (2015-02-13)
We have previously shown that underground railway particulate matter (PM) is rich in iron and other transition metals across coarse (PM10-2.5), fine (PM2.5), and quasi-ultrafine (PM0.18) fractions and is able to generate reactive oxygen species (ROS). However, there is little

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service