Skip to Content
Merck
All Photos(2)

Key Documents

203297

Sigma-Aldrich

Gadolinium(III) oxide

powder, ≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd2O3
CAS Number:
Molecular Weight:
362.50
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38161003
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: gadolinium

mp

2330 °C (lit.)

density

7.407 g/mL at 20 °C (lit.)

SMILES string

O=[Gd]O[Gd]=O

InChI

1S/2Gd.3O

InChI key

RNYFGGMJZRTZKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrée-Anne Guay-Bégin et al.
Langmuir : the ACS journal of surfaces and colloids, 28(1), 774-782 (2011-10-06)
The performance of nanomaterials for biomedical applications is highly dependent on the nature and the quality of surface coatings. In particular, the development of functionalized nanoparticles for magnetic resonance imaging (MRI) requires the grafting of hydrophilic, nonimmunogenic, and biocompatible polymers
Nikesh Gupta et al.
International journal of nanomedicine, 7, 5491-5500 (2012-12-13)
Mesoporous silica nanoparticles coencapsulating gadolinium oxide and horseradish peroxidase (HRP) have been synthesized in the aqueous core of sodium bis-(2-ethylhexyl) sulfosuccinate (AOT)-hexane-water reverse micelle. The average diameter of these silica particles is around 25 nm and the particles are spherical
Zijian Zhou et al.
Advanced materials (Deerfield Beach, Fla.), 24(46), 6223-6228 (2012-09-14)
Monodisperse Gd(2) O(3) -embedded iron oxide (GdIO) nanoparticles can simultaneously enhance the local magnetic field intensities of each other under an external magnetic field and result in synergistic enhancement of T(1) and T(2) effects. GdIO nanoparticles have the unique property
Eva Hemmer et al.
Acta biomaterialia, 9(1), 4734-4743 (2012-09-12)
Bioimaging is an important diagnostic tool in the investigation and visualization of biological phenomena in cells and in medicine. In this context, up-converting Gd(2)O(3):Er(3+),Yb(3+) nanostructures (nanoparticles, nanorods) have been synthesized by precipitation methods and hydrothermal synthesis. Independent of size and
Taejong Paik et al.
ACS nano, 7(3), 2850-2859 (2013-02-26)
Here, we report the shape-controlled synthesis of tripodal and triangular gadolinium oxide (Gd2O3) nanoplates. In the presence of lithium ions, the shape of the nanocrystals is readily controlled by tailoring reaction parameters such as temperature and time. We observe that

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service