Skip to Content
Merck
All Photos(1)

Key Documents

203009

Sigma-Aldrich

Cerium(III) sulfate octahydrate

99.999% trace metals basis

Synonym(s):

Cerium(3+) trisulfate octahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce2(SO4)3·8H2O
CAS Number:
Molecular Weight:
712.54
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤15.0 ppm Trace Metal Analysis

density

2.886 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.[Ce+3].[Ce+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Ce.3H2O4S.8H2O/c;;3*1-5(2,3)4;;;;;;;;/h;;3*(H2,1,2,3,4);8*1H2/q2*+3;;;;;;;;;;;/p-6

InChI key

PQBKXYUMEMUVIH-UHFFFAOYSA-H

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service