Skip to Content
Merck
All Photos(2)

Key Documents

200107

Sigma-Aldrich

Selenium dioxide

≥99.9% trace metals basis

Synonym(s):

NSC 56753, Selenium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SeO2
CAS Number:
Molecular Weight:
110.96
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 157 °C)

Assay

≥99.9% trace metals basis

form

solid

reaction suitability

reagent type: oxidant

mp

315 °C (subl.) (lit.)

SMILES string

O=[Se]=O

InChI

1S/O2Se/c1-3-2

InChI key

JPJALAQPGMAKDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Selenium dioxide (SeO2) can be used as:
  • An oxidizing agent to introduce carbonyl functional groups at activated carbon.
  • A catalyst in the allylic hydroxylation reactions.
  • An oxidant in the synthesis of amides by oxidative amidation of aldehydes with amines.
  • A catalyst to synthesize di and trioxidized bicyclic cyclopentenones by reacting with Pauson-Khand reaction (PKR) products.
  • A catalyst along with iodine (I2) to prepare mono and bis-sulfenylindoles by treating indoles with diaryl disulfides via C-H sulfenylation.
  • A catalyst to synthesize alkyl and aryl nitriles from the corresponding aldehydes via the formation of aldoxime.
  • A selenium precursor to synthesize FeSe nanostructures via electrochemical deposition method.

It can also be used as a versatile reagent for allylic oxidation and oxidative-elimination reactions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Bai et al.
Journal of nanoscience and nanotechnology, 11(6), 5012-5017 (2011-07-21)
This paper reported the preparation and antioxidant capacities of element selenium nanoparticles (nanoSe(0))-ascorbic acid (Vc) sol and nanSe(0)/Vc/selenocystine (SeCys) sol-gel compounds. NanoSe(0)-Vc sol was prepared by reduction of selenious dioxide (SeO2) with Vc. In the nanoSe(0)-Vc sol, highly concentrated Vc
Electrodeposition of nano-dimensioned FeSe
Chen PY, et al.
Thin Solid Films, 519(23), 8397-8400 (2011)
P. Y. Chen, et al.,
Thin Solid Films, 519, 8397-8400 (2011)
Selenium (IV) oxide
Hoekstra WJ, et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-12 (2001)
Selenium dioxide-promoted selective synthesis of mono-and bis-sulfenylindoles
Thurow S, et al.
Organic Chemistry Frontiers : An International Journal of Organic Chemistry / Royal Society of Chemistry, 5(12), 1983-1991 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service