Skip to Content
Merck
All Photos(1)

Documents

196894

Sigma-Aldrich

4-(Trifluoromethyl)benzoic acid

98%

Synonym(s):

α,α,α-Trifluoro-p-toluic acid, 4-Carboxybenzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CO2H
CAS Number:
Molecular Weight:
190.12
Beilstein:
2049241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

219-220 °C (lit.)

SMILES string

OC(=O)c1ccc(cc1)C(F)(F)F

InChI

1S/C8H5F3O2/c9-8(10,11)6-3-1-5(2-4-6)7(12)13/h1-4H,(H,12,13)

InChI key

SWKPKONEIZGROQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-(Trifluoromethyl)benzoic acid was used in the synthesis of salicylanilide 4-(trifluoromethyl)benzoates via N,N′-dicyclohexylcarbodiimide coupling in dry N,N-dimethylformamide. It was used as internal standard during the ultra trace analysis of fluorinated aromatic carboxylic acids by GC/MS method.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultra trace determination of fluorinated aromatic carboxylic acids in aqueous reservoir fluids by solid phase extraction in combination with negative ion chemical ionisation mass spectrometry after derivatisation with pentafluorobenzyl bromide.
Galdiga CU and Greibrokk T.
Fresenius Journal of Analytical Chemistry, 361(8), 797-802 (1998)
Total synthesis of the C-1027 chromophore core: extremely facile enediyne formation through Sml2-mediated 1,2-elimination.
Masayuki Inoue et al.
Angewandte Chemie (International ed. in English), 47(9), 1777-1779 (2008-01-24)
Fabio Ziarelli et al.
Journal of pharmaceutical and biomedical analysis, 59, 13-17 (2011-11-08)
This work shows that high resolution magic angle spinning (HR-MAS) can be used to elucidate ligand-receptor binding phenomena for mass-limited samples in liquid solution. With respect to conventional 5mm liquid-state NMR probe heads, HR-MAS allows for a considerable reduction (about
Martin Krátký et al.
Molecules (Basel, Switzerland), 17(8), 9426-9442 (2012-08-09)
Searching for novel antimicrobial agents still represents a current topic in medicinal chemistry. In this study, the synthesis and analytical data of eighteen salicylanilide esters with 4-(trifluoromethyl)benzoic acid are presented. They were assayed in vitro as potential antimycotic agents against
K H Engesser et al.
Archives of microbiology, 149(3), 198-206 (1988-01-01)
Enzymes of the p-cymene pathway in Pseudomonas putida strains cometabolized the intermediate analogue 4-trifluoromethyl(TFM)benzoate. Three products, 4-TFM-2,3-dihydro-2,3-dihydroxybenzoate, 4-TFM-2,3-dihydroxybenzoate and 2-hydroxy-6-oxo-7,7,7-trifluorohepta-2,4-dienoate (7-TFHOD) were identified chemically and by spectroscopic properties. Certain TFM-substituted analogue metabolites of the p-cymene pathway were transformed at drastically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service