Skip to Content
Merck
All Photos(1)

Documents

196274

Sigma-Aldrich

Thiomorpholine

98%

Synonym(s):

Tetrahydro-2H-1,4-thiazine, Thiamorpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NS
CAS Number:
Molecular Weight:
103.19
Beilstein:
102550
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.538 (lit.)

bp

169 °C (lit.)

solubility

organic solvents: miscible(lit.)
water: miscible(lit.)

density

1.026 g/mL at 25 °C (lit.)

SMILES string

C1CSCCN1

InChI

1S/C4H9NS/c1-3-6-4-2-5-1/h5H,1-4H2

InChI key

BRNULMACUQOKMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thiomorpholine forms complexes with Cu(II), Pt(II) and Ni(II) salts and their catalytic activity has been investigated.

Application

Thiomorpholine has been used in the preparation of:
  • N-Boc-α-alkyl-β-(sec-amino)alanines
  • thiomorpholine-N-borane

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesut Kacan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 118, 572-577 (2013-10-05)
Several Cu(II), Pt(II) and Ni(II) complexes of N-substituted, piperazine (NN donor), morpholine (NO donor) and thiomorpholine (NS donor) derivatives were synthesized and their thermal behavior and catalytic activity in epoxidation reaction of cis-diphenylethylene were studied using oxygen sources NaOCl. The
Synthesis and comparative reactivity of thiomorpholine-borane: aqueous hydrolysis and oxidation by hypochlorite.
Amezcua CA, et al.
Inorgorganica Chimica Acta, 290(1), 80-85 (1999)
Amany Belal
Bioorganic chemistry, 59, 124-129 (2015-03-10)
A new series of pyrrolizine derivatives 4-8c were synthesized, their structures were confirmed by spectral and elemental analyses. Cytotoxic activity of these compounds was evaluated against breast (MCF7), colon (HCT116) and liver (HEPG2) cancer cell lines using sulphorhodamine-B (SRB) assay
B Beck et al.
Molecular diversity, 14(3), 479-491 (2010-04-22)
We designed two novel thiolactone scaffolds. Both scaffolds can be accessed by a convergent Ugi multicomponent reaction (MCR) and are, thus, amenable to library synthesis. Design, stereoselectivity, structures, full experimental details, and virtual libraries will be reported.
B Combourieu et al.
Biodegradation, 9(6), 433-442 (1999-05-21)
Spectrophotometric assays of Mycobacterium aurum MO1 cells extracts gave evidence of a soluble cytochrome P450, involved in the degradative pathway of morpholine, a waste product from the chemical industry. In order to get further information, the kinetics of the biodegradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service