Skip to Content
Merck
All Photos(2)

Key Documents

192392

Sigma-Aldrich

2-Chloropropionamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(Cl)CONH2
CAS Number:
Molecular Weight:
107.54
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

77-81 °C (lit.)

SMILES string

CC(Cl)C(N)=O

InChI

1S/C3H6ClNO/c1-2(4)3(5)6/h2H,1H3,(H2,5,6)

InChI key

OEZPDHRXGCLGKB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y F Li et al.
Journal of biochemistry, 124(1), 20-22 (1998-06-30)
The crystal structure of a complex prepared by soaking a single crystal of the S175A mutant of L-2-haloacid dehalogenase from Pseudomonas sp. YL in a solution containing a poor substrate, L-2-chloropropionamide, has been determined by X-ray analysis at 2. 15
S J Hope et al.
Archives of microbiology, 163(1), 57-64 (1995-01-01)
Mutants of two strains of Pseudomonas putida expressed two cryptic chloroamidases (C-amidase and H-amidase) and one cryptic dehalogenase (DehII). The mutants were selected on either 2-chloropropionamide (2CPA) or 2-monochloropropionate (2MCPA), developing as papillae in parental colonies growing on a metabolisable
Synthesis of zwitterionic polymer by SET-LRP at room temperature in aqueous.
Ding W, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 49(2), 432-440 (2011)
Controled/Living Radical Polymerization of Acrylamide in Aqueous Media by ATRP.
AN H-Y and SONG C-L.
Journal of Petrochemical Universities / Shi You Hua Gong Gao Deng Xue Xiao Xue Bao, 2, 007-007 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service