Skip to Content
Merck
All Photos(1)

Key Documents

192171

Sigma-Aldrich

DL-Alaninol

98%

Synonym(s):

(±)-2-Amino-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)CH2OH
CAS Number:
Molecular Weight:
75.11
Beilstein:
1209234
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

optical activity

[α]/D +1.0 to −1.0°

refractive index

n20/D 1.450 (lit.)

bp

173-176 °C (lit.)

density

0.943 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CC(N)CO

InChI

1S/C3H9NO/c1-3(4)2-5/h3,5H,2,4H2,1H3

InChI key

BKMMTJMQCTUHRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

DL-Alaninol has been used in the synthesis of:
  • N6-α-(I-hydroxypropyl) lysine
  • diastereomers of DL-β-amino alcohols
  • enantiopure and racemic samples of 2-methyl-N-tosylaziridine
  • (±)-3-(5-dimethylcarbamoyl-pent-1-enyl)-N-(2-hydroxy-1-methyl-ethyl)benzamide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian C Stewart et al.
Journal of the American Chemical Society, 127(50), 17616-17617 (2005-12-15)
Sulfonylaziridines have been identified as excellent monomers for living ring-opening polymerization initiated by nucleophilic sulfonylamides. The resulting polymers exhibit low polydispersities and controllable molecular weights. The enantiopurity of the monomer plays a key role: racemic samples yield soluble polymers of
P M Hoi et al.
British journal of pharmacology, 152(5), 751-764 (2007-09-25)
A putative novel cannabinoid receptor mediates vasorelaxation to anandamide and abnormal-cannabidiol and is blocked by O-1918 and by high concentrations of rimonabant. This study investigates VSN16, a novel water-soluble agonist, as a vasorelaxant potentially acting at non-CB1, non-CB2 cannabinoid receptors
The role of lysine residues in the activity of 2-keto-3-deoxy-6-phosphogluconate aldolase.
J M Ingram et al.
The Journal of biological chemistry, 240(11), 4146-4151 (1965-11-01)
The substrate-dependent steric course of the ethanolamine ammonia-lyase reaction.
P Diziol et al.
European journal of biochemistry, 106(1), 211-224 (1980-05-01)
Julien Pytkowicz et al.
Organic & biomolecular chemistry, 8(20), 4540-4542 (2010-09-08)
Two efficient routes are reported for the synthesis of both enantiomers of trifluoroalaninol in enantiopure form. The first pathway involves a Strecker-type reaction performed from a chiral trifluoromethyloxazolidine (Fox). The second route, which is more direct, involves, as a key

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service