Skip to Content
Merck

190837

Poly(1-vinylpyrrolidone-co-vinyl acetate) solution

average Mw ~13,000 (GPC vs. poly(ethylene oxide)), 50 wt. % in isopropanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mol wt

average Mw ~13,000 (GPC vs. poly(ethylene oxide))

concentration

50 wt. % in isopropanol

density

0.958 g/mL at 25 °C

SMILES string

N1(CCCC1=O)C=C.O(C(=O)C)C=C

InChI

1S/C6H9NO.C4H6O2/c1-2-7-5-3-4-6(7)8;1-3-6-4(2)5/h2H,1,3-5H2;3H,1H2,2H3

InChI key

FYUWIEKAVLOHSE-UHFFFAOYSA-N

General description

∼1:2.4 mole ratio of VP:VA


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

66.2 °F

flash_point_c

19 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library