188832
15-Crown-5
98%
Synonym(s):
15C5, 1,4,7,10,13-Pentaoxacyclopentadecane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H20O5
CAS Number:
Molecular Weight:
220.26
Beilstein:
1618144
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.465 (lit.)
bp
93-96 °C/0.05 mmHg (lit.)
density
1.113 g/mL at 20 °C (lit.)
functional group
ether
SMILES string
O1CCOCCOCCOCCOCC1
InChI
1S/C10H20O5/c1-2-12-5-6-14-9-10-15-8-7-13-4-3-11-1/h1-10H2
InChI key
VFTFKUDGYRBSAL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
15-Crown-5 is a crown ether generally used as a ligand in coordination chemistry because of its strong chelating property with certain alkali cations to form complexes. Some derivatives of 15-crown-5 are used as sensors and probes in different physical-chemical processes, phase-transfer reactions, and selective capture of ions for separation and transport.
Application
15-Crown-5 can be used:
- As a phase transfer catalyst for the preparation of polyphenylacetylene (PPA) by anionic polymerization of phenylacetylene with sodium amide (NaNH2).
- In the allylic alkylation of allyl acetates by malonates with palladium complex catalysts.
15-Crown-5 is a crown ether that is generally used as a ligand in coordination chemistry. It shows the ability to complexes with alkali metal ions.
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Crown ethers for chemical analysis: A Review.
Yoshio M, et al.
Analytical Letters, 15(15), 1197-1276 (1982)
Mechanistic and synthetic studies in catalytic allylic alkylation with palladium complexes of 1-(2-diphenylphosphino-1-naphthyl) isoquinoline
Brown JM, et al.
Tetrahedron, 50(15), 4493-4506 (1994)
A PVC-based benzo-15-crown-5 membrane sensor for cadmium
Srivastava SK, et al.
Electroanalysis, 8(10), 938-940 (1996)
Novel benzo-15-crown-5 sol-gel coating for solid-phase microextraction
Wang D, et al.
Journal of Chromatography A, 1005(1-2), 1-12 (2003)
15-Crown-5.
Parsons S, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(7), o3130-o3130 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service