Skip to Content
Merck
All Photos(1)

Key Documents

186988

Sigma-Aldrich

2-Ethylfuran

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O
CAS Number:
Molecular Weight:
96.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

92-93 °C/768 mmHg (lit.)

density

0.912 g/mL at 25 °C (lit.)

SMILES string

CCc1ccco1

InChI

1S/C6H8O/c1-2-6-4-3-5-7-6/h3-5H,2H2,1H3

InChI key

HLPIHRDZBHXTFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylfuran undergoes tetraphenylporphin-photosensitized oxygenation in non-polar aprotic solvents via (4+2)-cycloaddition of singlet oxygen to yield the corresponding monomeric unsaturated secondary ozonide.

Application

2-Ethylfuran was used to investigate the gas phase products formed from the Cl atoms initiated reactions of 2-ethylfuran by in situ long-path FTIR absorption spectroscopy. It was also used in the synthesis of 4-oxo-(E)-2-hexenal via ring opening reaction using aqueous N-bromosuccinimide.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atmospheric degradation of alkylfurans with chlorine atoms: Product and mechanistic study.
Villanueva F, et al.
Atmospheric Environment, 43(17), 2804-2813 (2009)
Singlet oxygen photooxygenation of furans: Isolation and reactions of (4+ 2)-cycloaddition products (unsaturated sec.-ozonides).
Gollnick K and Griesbeck A.
Tetrahedron, 41(11), 2057-2068 (1985)
Koji Noge et al.
Journal of chemical ecology, 38(8), 1050-1056 (2012-10-12)
We examined whether shared volatiles found in various heteropteran species and developmental stages function to repel predators. The nymphal dorsal abdominal gland secretions of Riptortus pedestris (Heteroptera: Alydidae) and Thasus acutangulus (Heteroptera: Coreidae), and the metathoracic scent gland secretion of
Francisco J Hidalgo et al.
Journal of agricultural and food chemistry, 62(49), 12045-12051 (2014-11-25)
The carbonyl-scavenging ability of 2-amino-1-methylbenzimidazole (AMBI) and the heterocyclic aromatic amine 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) was investigated in an attempt to identify new routes that can modify the carbonyl content of foods. The reaction of both AMBI and PhIP with 2-alkenals, 2,4-alkadienals
Audrey R Smith et al.
Journal of mass spectrometry : JMS, 50(11), 1206-1213 (2015-10-28)
Absolute photoionization cross sections of the molecules 2-ethylfuran, 2-acetylfuran and furfural, including partial ionization cross sections for the dissociative ionized fragments, are measured for the first time. These measurements are important because they allow fuel quantification via photoionization mass spectrometry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service