Skip to Content
Merck
All Photos(3)

Documents

185574

Sigma-Aldrich

1,1,2,2-Tetrabromoethane

98%

Synonym(s):

Acetylene tetrabromide, Muthmanns liquid, TBE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2CHCHBr2
CAS Number:
Molecular Weight:
345.65
Beilstein:
1098321
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

11.9 (vs air)

Quality Level

vapor pressure

0.1 mmHg ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

635 °F

refractive index

n20/D 1.637 (lit.)

bp

119 °C/15 mmHg (lit.)

mp

−1-1 °C (lit.)

solubility

alcohol: miscible
chloroform: miscible
diethyl ether: miscible
glacial acetic acid: miscible
water: insoluble

density

2.967 g/mL at 25 °C (lit.)

SMILES string

BrC(Br)C(Br)Br

InChI

1S/C2H2Br4/c3-1(4)2(5)6/h1-2H

InChI key

QXSZNDIIPUOQMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Density and viscosity of mixtures of 1,1,2,2-tetrabromoethane and 1-bromododecane has been measured at atmospheric pressure.

Application

1,1,2,2-Tetrabromoethane was used to prepare tribromoethene by dehydrohalogenation using sodium hydroxide in methanol.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Density and viscosity of mixtures of 1, 1, 2, 2-tetrabromoethane and 1-bromododecane.
Cokelet GR, et al.
Journal of Chemical and Engineering Data, 14(4), 470-473 (1969)
J K Magnuson et al.
Applied and environmental microbiology, 66(12), 5141-5147 (2000-12-01)
The anaerobic bacterium Dehalococcoides ethenogenes is the only known organism that can completely dechlorinate tetrachloroethene or trichloroethene (TCE) to ethene via dehalorespiration. One of two corrinoid-containing enzymes responsible for this pathway, TCE reductive dehalogenase (TCE-RDase) catalyzes the dechlorination of TCE
Zhaojiang Guo et al.
Journal of invertebrate pathology, 126, 21-30 (2015-01-18)
The Gram-positive bacterium Bacillus thuringiensis (Bt) produces Cry toxins that have been used to control important agricultural pests. Evolution of resistance in target pests threatens the effectiveness of these toxins when used either in sprayed biopesticides or in Bt transgenic
R Galuppi et al.
Veterinary parasitology, 210(1-2), 10-18 (2015-04-15)
The present study aims to evaluate the prevalence, pattern of spread and risk factors for the transmission of cryptosporidiosis in foals and mares hospitalized in a University Equine Perinatology Unit, where a new subtype family of Cryptosporidium horse genotype was
Somin Eunice Lee et al.
Nano letters, 15(7), 4564-4570 (2015-06-04)
Plasmon rulers, consisting of pairs of gold nanoparticles, allow single-molecule analysis without photobleaching or blinking; however, current plasmon rulers are irreversible, restricting detection to only single events. Here, we present a reversible plasmon ruler, comprised of coupled gold nanoparticles linked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service