Skip to Content
Merck
All Photos(3)

Key Documents

182354

Sigma-Aldrich

Poly(1,4-phenylene sulfide) average Mn ~ 10,000, powder | 25212-74-2

average Mn ~10,000, powder

Synonym(s):

Poly(1,4-Phenylene Sulfide)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

average Mn ~10,000

mp

285-300 °C

transition temp

Tg 150 °C

density

1.36 g/mL at 25 °C

General description

Poly(1,4-phenylene sulfide)(PPS) is an aromatic polymer that can be used as a polymer backbone or an engineering plastic. It is also called as poly(phenylene sulfide) which can be sulfonated to form ion-exchange membranes for electrochemical applications. It is industrially produced by polycondensation of p-dichlorobenzene and sodium sulfate in N-methylpyrrolidone as a solvent.

Application

PPS can be sulfonated to form a proton exchange membrane with zeolite that can be used as an electrolyte with higher thermal stability which facilitates in the fabrication of solar cells. It can also be used in the formation of organic- inorganic nanocomposite membranes using nanozeolites and carbon nanotubes as nanofillers which can be used as high performance gas separation membranes. PPS may be sulfonated and chloromethylated to form novel PPS fiber with higher dyeing ability in dispersive, anionic and cationic dyes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystallization kinetics of polyphenylene sulfide
Jog, J. P., & Nadkarni, V. M.
Journal of Applied Polymer Science, 30(3), 997-1009 (1985)
Crystallization, morphology, and thermal behavior of poly (p-phenylene sulfide).
Silvestre, C., Di Pace, E., Napolitano, R., Pirozzi, B., & Cesario, G.
Journal of Polymer Science. Part B, Polymer Physics, 39(4), 415-424 (2001)
An optical study of the arsenic pentafluoride doping of poly (p-phenylene sulphide): polaron and bipolaron transitions
Friend, R. H., & Giles, J. R.
Journal of the Chemical Society. Chemical Communications, 16, 1101-1103 (1984)
Design of poly (1-hexadecene-sulfone)/poly (1, 4-phenylene sulfide) membrane containing nano-zeolite and carbon nanotube for gas separation.
Kausar A.
International Journal of Plastics Technology, 21(1), 96-107 (2017)
Polymerization of 4-Bromobenzenethiol to Poly (1, 4-phenylene sulfide) with a Free Radical Initiator.
Ding Y and Hay AS
Macromolecules, 30(6), 1849-1850 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service