Skip to Content
Merck
All Photos(4)

Key Documents

181978

Sigma-Aldrich

Poly(ethyleneimine) solution

average Mn ~60,000 by GPC, average Mw ~750,000 by LS, 50 wt. % in H2O

Synonym(s):

Ethyleneimine polymer solution, PEI

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

9 mmHg ( 20 °C)

Quality Level

mol wt

average Mn ~60,000 by GPC
average Mw ~750,000 by LS

concentration

50 wt. % in H2O

SMILES string

N1CC1

InChI

1S/C2H5N/c1-2-3-1/h3H,1-2H2

InChI key

NOWKCMXCCJGMRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Detergents, adhesives, water treatment, printing inks, dyes, cosmetics, and paper industry, adhesion promoter, lamination primer, fixative agent, flocculant, cationic dispersant, stability enhancer, surface activator, chelating agent, scavenger for aldehydes and oxides.
Protein precipitant.[1]

Physical form

Branched polymer

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Y Zhao et al.
Journal of biotechnology, 14(3-4), 273-283 (1990-06-01)
Protein recovery from industrial microbial processes can be very expensive, often exceeding the cost of protein production. We have genetically engineered 3 beta-galactosidase (beta-gal) fusion proteins containing poly-aspartic acid tails to test the effect of the tails on recovery by
Andrew Jajack et al.
PloS one, 14(1), e0210286-e0210286 (2019-01-17)
Insurmountable detection challenges will impede the development of many of the next-generation of lab-on-a-chip devices (e.g., point-of-care and real-time health monitors). Here we present the first membrane-based, microfluidic sample preconcentration method that is continuous, quantifiable, simple, and capable of working
Jason C Casler et al.
Molecular biology of the cell, 31(26), 2892-2903 (2020-10-29)
Membrane traffic can be studied by imaging a cargo protein as it transits the secretory pathway. The best tools for this purpose initially block export of the secretory cargo from the endoplasmic reticulum (ER) and then release the block to
Xinyue Yuan et al.
Nature communications, 11(1), 4854-4854 (2020-09-27)
Chronic imaging of neuronal networks in vitro has provided fundamental insights into mechanisms underlying neuronal function. Current labeling and optical imaging methods, however, cannot be used for continuous and long-term recordings of the dynamics and evolution of neuronal networks, as
Kevin Bellande et al.
Plant physiology and biochemistry : PPB, 157, 441-452 (2020-11-20)
An efficient purification of recombinant proteins often requires a high ratio of recombinant to host proteins. In plants, Ribulose-1,5-bisphosphate carboxylase/oxygenase (RuBisCO) is the most abundant leaf protein, thus strongly impacting purification yield. Here, we describe a simple and robust purification

Articles

Gene therapy has become one of the most discussed techniques in biomedical research in recent years.

Professor Yoshiki Katayama (Kyushu University, Japan) discusses recent advances in drug delivery systems and strategies that exploit the EPR effect, with a special focus on stimuli-responsive systems based on novel materials.

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Microfluidic assembly improves polyamine nanoencapsulation of nucleic acids, overcoming challenges like polydispersity and poor reproducibility.

See All

Questions

1–8 of 8 Questions  
  1. Are there any details available about the amine group distribution of this polyethyleneimine?

    1 answer
    1. Unfortunately, the amine group distribution in this product is not determined or provided in the product documentation. This means that information about the exact ratio of primary, secondary, and tertiary amines in this particular polyethyleneimine may not be available.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  4. I don't see storage information here. What are the storage recommendations for this product?

    1 answer
    1. This product should be stored in room temperature. Store under inert gas. Please note that this chemical is sensitive to carbon dioxide.

      Helpful?

  5. Product 181978, Poly(ethyleneimine) solution, is listed as a solution but it is sold by weight; is this a solid or a liquid?

    1 answer
    1. This product is a solution of Poly(ethyleneimine) in water. However, this product, like many Sigma-Aldrich products, is weighed into the bottle. If 5 grams of this solution is purchased, approximately 4.62mL will be received.

      Helpful?

  6. What possible impurities are present in Product 181978, Poly(ethyleneimine) solution?

    1 answer
    1. Although Sigma-Aldrich does not currently test for the impurities in this product, 2-aminoethanol and less than 50 ppb of unreacted monomer may be present as a result of the manufacturing process.

      Helpful?

  7. I see two molecular weight values reported for Product 181978, Poly(ethyleneimine) solution. Which one is correct?

    1 answer
    1. Both of the approximate molecular weights listed for this product are correct based on the testing methods used by Sigma-Aldrich. However, each molecular weight is determined by a different method. The Mn refers to the number-average molecular weight (determined by GPC) while the Mw refers to the weight-average molecular weight (determined by Light Scattering). These two numbers are different but still related. The polydispersity is the measure of uniformity of the molecules in a polymer sample. A ratio of the weight-average molecular and the number-average molecular weight indicates how uniform the polymer sample is. The greater the ratio the less uniform the polymer molecules.

      Helpful?

  8. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service