178535
N,N-Dimethylformamide diisopropyl acetal
95%
Synonym(s):
1,1-Diisopropoxy-N,N-dimethylmethylamine, 1,1-Diisopropoxytrimethylamine
About This Item
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.401 (lit.)
bp
79-80 °C/60 mmHg (lit.)
density
0.838 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CC(C)OC(OC(C)C)N(C)C
InChI
1S/C9H21NO2/c1-7(2)11-9(10(5)6)12-8(3)4/h7-9H,1-6H3
InChI key
XOZZATXWQMOVHL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
77.0 °F - closed cup
Flash Point(C)
25 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service