Skip to Content
Merck
All Photos(1)

Documents

176087

Sigma-Aldrich

Urea-d4

98 atom % D

Synonym(s):

Carbamide-d4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D2NCOND2
CAS Number:
Molecular Weight:
64.08
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

form

solid

technique(s)

NMR: suitable

mp

132-135 °C (lit.)

mass shift

M+4

SMILES string

[2H]N([2H])C(=O)N([2H])[2H]

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)/i/hD4

InChI key

XSQUKJJJFZCRTK-JBISRTOLSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

C F Araujo et al.
Physical chemistry chemical physics : PCCP, 19(27), 17998-18009 (2017-07-01)
The solids choline chloride and urea, mixed in a 1 : 2 molar proportion, form the iconic deep eutectic solvent "Reline". A combination of computational and vibrational spectroscopy tools, including inelastic neutron scattering (INS), have been used to probe intermolecular interactions in
Alexandra Tsirigotaki et al.
Structure (London, England : 1993), 26(5), 695-707 (2018-04-03)
Secretory preproteins carry signal peptides fused amino-terminally to mature domains. They are post-translationally targeted to cross the plasma membrane in non-folded states with the help of translocases, and fold only at their final destinations. The mechanism of this process of
Olivier Julien et al.
Protein science : a publication of the Protein Society, 18(10), 2172-2182 (2009-08-21)
Prion diseases, or transmissible spongiform encephalopathies, are a group of infectious neurological diseases associated with the structural conversion of an endogenous protein (PrP) in the central nervous system. There are two major forms of this protein: the native and noninfectious

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service