Skip to Content
Merck
All Photos(1)

Key Documents

175900

Sigma-Aldrich

o-Xylene-d10

99 atom % D

Synonym(s):

1,2-Dimethylbenzene-d10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D4(CD3)2
CAS Number:
Molecular Weight:
116.23
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

liquid

technique(s)

GC/MS: suitable
NMR: suitable

refractive index

n20/D 1.5016 (lit.)

bp

142 °C (lit.)

mp

-25 °C (lit.)

density

0.953 g/mL at 25 °C (lit.)

mass shift

M+10

SMILES string

[2H]c1c([2H])c([2H])c(c(c1[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C8H10/c1-7-5-3-4-6-8(7)2/h3-6H,1-2H3/i1D3,2D3,3D,4D,5D,6D

InChI key

CTQNGGLPUBDAKN-ZGYYUIRESA-N

General description

o-Xylene-d10 is a deuterated derivative of o-xylene. It has an isotopic purity of 99atom%D. It participates as an internal standard during the quantification of volatile organic compounds (furan, chloroform, benzene, trichloroethene, toluene and styrene) by vacuum distillation coupled with gas chromatography/mass spectrometry.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia J Nyman et al.
Journal of AOAC International, 97(2), 510-520 (2014-05-17)
Recent studies showed that headspace and purge and trap methods have limitations when used to determine volatile organic compounds (VOCs) in foods, including matrix effects and artifact formation from precursors present in the sample matrix or from thermal decomposition. U.S.
Poguang Wang et al.
Journal of the American Society for Mass Spectrometry, 26(10), 1713-1721 (2015-06-28)
N-(2-(Bromomethyl)benzyl)-N,N-diethylethanaminium bromide, that we designate as CAX-B (cationic xylyl-bromide), is presented as a derivatization reagent for increasing sensitivity in mass spectrometry. Because of its aryl bromomethyl moiety, CAX-B readily labels compounds having an active hydrogen. In part, a CAX-tagged analyte

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service