Recommended Products
Assay
98%
form
fibers
mp
146-148 °C (lit.)
solubility
acetone: soluble 2.5%, clear to slightly hazy, yellow
functional group
nitro
SMILES string
[O-][N+](=O)c1ccc2OCOc2c1
InChI
1S/C7H5NO4/c9-8(10)5-1-2-6-7(3-5)12-4-11-6/h1-3H,4H2
InChI key
SNWQAKNKGGOVMO-UHFFFAOYSA-N
General description
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
289.4 °F - closed cup
Flash Point(C)
143 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service