All Photos(3)
About This Item
Linear Formula:
CH3C6H4COCH2Br
CAS Number:
Molecular Weight:
213.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
90%
bp
105 °C/0.1 mmHg (lit.)
mp
45-49 °C (lit.)
functional group
bromo
ketone
storage temp.
2-8°C
SMILES string
Cc1ccc(cc1)C(=O)CBr
InChI
1S/C9H9BrO/c1-7-2-4-8(5-3-7)9(11)6-10/h2-5H,6H2,1H3
InChI key
KRVGXFREOJHJAX-UHFFFAOYSA-N
Related Categories
General description
2-Bromo-4′-methylacetophenone is an α-bromoketone.
Application
2-Bromo-4′-methylacetophenone was used in the general fluorous thiol quenching method. It was also used in the preparation of hydroxyquinolinone and N-derivatized carboxamides.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Miroslav Soural et al.
Journal of combinatorial chemistry, 9(5), 793-796 (2007-08-07)
A highly efficient solid-phase synthesis of 2-substituted-3-hydroxy-4(1H)-quinolinone-7-carboxamides was developed using anthranilates and bromoketones as the key synthons. Primary amines immobilized to an acid-cleavable BAL linker were acylated with 1-methyl-2-aminoterephtalate. After cleavage of the methyl ester, bromoketones were used to form
Use of fluorous silica gel to separate fluorous thiol quenching derivatives in solution-phase parallel synthesis
Zhang W, et al.
Tetrahedron, 58(20), 3871-3875 (2002)
Hana Elshaflu et al.
Frontiers in chemistry, 6, 247-247 (2018-07-19)
The novel approach in the treatment of complex multifactorial diseases, such as neurodegenerative disorders and cancer, requires a development of efficient multi-targeting oriented drugs. Since oxidative stress significantly contributes to the pathogenesis of cancer and neurodegenerative disorders, potential drug candidates
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service