Skip to Content
Merck
All Photos(2)

Key Documents

156299

Sigma-Aldrich

2-Chloroanthraquinone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H7ClO2
CAS Number:
Molecular Weight:
242.66
Beilstein:
2051842
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

209-211 °C (lit.)

functional group

chloro
ketone

SMILES string

Clc1ccc2C(=O)c3ccccc3C(=O)c2c1

InChI

1S/C14H7ClO2/c15-8-5-6-11-12(7-8)14(17)10-4-2-1-3-9(10)13(11)16/h1-7H

InChI key

FPKCTSIVDAWGFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

This product has been evaluated as a photo-reagent for the analysis of ginsenosides using photoreduction fluorescence (PRF) detection method.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoreactivity of anthraquinones for the analysis of ginsenosides using photoreduction fluorescence detection-HPLC
Park MK, et al.
Archives of Pharmacal Research, 19(6), 562-565 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service