15502
1-Boc-piperazine
≥98.0% (GC)
Synonym(s):
tert-Butyl piperazine-1-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H18N2O2
CAS Number:
Molecular Weight:
186.25
Beilstein:
879985
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Quality Level
Assay
≥98.0% (GC)
form
solid
mp
43-47 °C (lit.)
43-47 °C
SMILES string
CC(C)(C)OC(=O)N1CCNCC1
InChI
1S/C9H18N2O2/c1-9(2,3)13-8(12)11-6-4-10-5-7-11/h10H,4-7H2,1-3H3
InChI key
CWXPZXBSDSIRCS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
1-Boc-piperazine undergoes Buchwald-Hartwig amination with various aryl halides to form corresponding amine derivatives. It can be used to synthesize monosubstituted piperazine intermediates of many bioactive molecules and piperazine containing drug substances, such as trazodone.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and biological evaluation of new 2-(6-alkyl-pyrazin-2-yl)-1H-benz [d] imidazoles as potent anti-inflammatory and antioxidant agents.
Shankar B, et al.
Medicinal Chemistry Research, 26(9), 1835-1846 (2017)
Synthesis and dual D2 and 5-HT1A receptor binding affinities of 5-piperidinyl and 5-piperazinyl-1 H-benzo [d] imidazol-2 (3 H)-ones.
Ullah N
Journal of Enzyme Inhibition and Medicinal Chemistry, 29(2), 281-291 (2014)
Conformationally rigid derivatives of WAY-267,464: Synthesis and pharmacology at the human oxytocin and vasopressin-1a receptors.
Jorgensen WT, et al.
European Journal of Medicinal Chemistry, 143, 1644-1656 (2018)
Discovery of 1-[2-(2, 4-dimethylphenylsulfanyl) phenyl] piperazine (Lu AA21004): a novel multimodal compound for the treatment of major depressive disorder.
Bang-Andersen B, et al.
Journal of Medicinal Chemistry, 54(9), 3206-3221 (2011)
Nisar Ullah
Journal of enzyme inhibition and medicinal chemistry, 29(2), 281-291 (2013-03-16)
A series of new 5-piperidinyl and 5-piperazinyl-1H-benzo[d]imidazol-2(3H)-ones have been synthesized and evaluated for dual D2 and 5-HT1A receptor binding affinities. The synthesized ligands are structurally related to bifeprunox, a potential atypical antipsychotic, having potent D2 receptor antagonist and 5-HT1A receptor
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service