Skip to Content
Merck
All Photos(1)

Key Documents

15434

Sigma-Aldrich

Boc-Tyr(tBu)-OH

≥99.0% (sum of enantiomers, TLC)

Synonym(s):

Boc-O-tert-butyl-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COC6H4CH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
337.41
Beilstein:
4327336
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (sum of enantiomers, TLC)

optical activity

[α]20/D −15.0±1.0°, c = 1% in DMF

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1ccc(OC(C)(C)C)cc1)C(O)=O

InChI

1S/C18H27NO5/c1-17(2,3)23-13-9-7-12(8-10-13)11-14(15(20)21)19-16(22)24-18(4,5)6/h7-10,14H,11H2,1-6H3,(H,19,22)(H,20,21)/t14-/m0/s1

InChI key

ZEQLLMOXFVKKCN-AWEZNQCLSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ria Schönauer et al.
Journal of peptide science : an official publication of the European Peptide Society, 21(12), 905-912 (2016-01-16)
The human adrenomedullin (ADM) is a 52 amino acid peptide hormone belonging to the calcitonin family of peptides, which plays a major role in the development and regulation of cardiovascular and lymphatic systems. For potential use in clinical applications, we
Genevra Kuziel et al.
Cancers, 12(8) (2020-08-01)
Obesity is correlated with breast tumor desmoplasia, leading to diminished chemotherapy response and disease-free survival. Obesity causes chronic, macrophage-driven inflammation within breast tissue, initiated by chemokine ligand 2 (CCL2) signaling from adipose stromal cells. To understand how CCL2-induced inflammation alters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service