153079
3-Nitrobenzotrifluoride
97%
Synonym(s):
3-Nitro-α,α,α-trifluorotoluene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
O2NC6H4CF3
CAS Number:
Molecular Weight:
191.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.472 (lit.)
bp
200-205 °C (lit.)
density
1.436 g/mL at 25 °C (lit.)
functional group
fluoro
nitro
SMILES string
[O-][N+](=O)c1cccc(c1)C(F)(F)F
InChI
1S/C7H4F3NO2/c8-7(9,10)5-2-1-3-6(4-5)11(12)13/h1-4H
InChI key
WHNAMGUAXHGCHH-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
190.4 °F - closed cup
Flash Point(C)
88 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
H V Lavrov et al.
Dalton transactions (Cambridge, England : 2003), 46(33), 10926-10934 (2017-08-03)
We have predicted earlier by DFT simulation that tridentate O,N,O-donor cyclic dilactams (B) belonging to the family of pyridine-2,6-dicarboxamides are much more selective and efficient extractants for the separation of lanthanides and actinides than open-structure pyridine-2,6-dicarboxamides due to the higher
The Structures of Biphenyl, o-Terphenyl and Tetraphenylene1.
Karle IL and Brockway LO.
Journal of the American Chemical Society, 66(11), 1974-1979 (1944)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service