Skip to Content
Merck
All Photos(1)

Key Documents

15068

Sigma-Aldrich

Ethylenediamine solution

technical, 75-80%

Synonym(s):

1,2-Diaminoethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NH2
CAS Number:
Molecular Weight:
60.10
Beilstein:
4123523
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39030201
PubChem Substance ID:
NACRES:
NA.21

grade

technical

Assay

75-80%

form

liquid

SMILES string

NCCN

InChI

1S/C2H8N2/c3-1-2-4/h1-4H2

InChI key

PIICEJLVQHRZGT-UHFFFAOYSA-N

Gene Information

human ... FNTA(2339)

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediamine solution is a valuable reagent that can be used in a variety of organic synthesis reactions. It is a versatile solvent, a strong nucleophile, and a catalyst. It is also used as a chelating agent to bind metal ions in solution. This can be useful for a variety of applications, including metal recovery, wastewater treatment, and analytical chemistry.

Application

Ethylenediamine solution can be used as a building block for the synthesis of multi-amine dendritic poly(amidoamine) macromolecules. It is also used as a functionalization agent in cubic ZIF-8 metal-organic framework synthesis. Additionally, ethylenediamine is used for amide functionalization of MWNT/nylon 6, 6 composite nanofibers fabrication.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Zuidema
Pharmaceutisch weekblad. Scientific edition, 7(4), 134-140 (1985-08-23)
Ethylenediamine is an excipient with many industrial and pharmaceutical uses. It is included in creams as a stabilizer and in aminophylline as the counter ion of theophylline. Ethylenediamine is one of the most frequent contact sensitizers, producing local and generalized
Maria V Babak et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(13), 4308-4318 (2013-01-24)
With the aim of systematically studying fundamental structure-activity relationships as a basis for the development of Ru(II) arene complexes (arene = p-cymene or biphenyl) bearing mono-, bi-, or tridentate am(m)ine ligands as anticancer agents, a series of ammine, ethylenediamine, and
Ana Carolina M S Dias et al.
Biosensors & bioelectronics, 44, 216-221 (2013-02-23)
An immunosensor for the non-structural protein 1 (NS1) of the dengue virus based on carbon nanotube-screen printed electrodes (CNT-SPE) was successfully developed. A homogeneous mixture containing carboxylated carbon nanotubes was dispersed in carbon ink to prepare a screen printed working
Kavita Sablok et al.
Journal of hazardous materials, 248-249, 322-328 (2013-02-19)
Binding of electron-deficient trinitrotoluene (TNT) to the electron rich amine groups on a substrate form specific charge-transfer Jackson-Meisenheimer (JM) complex. In the present work, we report formation of specific JM complex on amine functionalized reduced graphene oxide/carbon nanotubes- (a-rGO/CNT) nanocomposite
Changhai Xu et al.
Carbohydrate polymers, 92(1), 249-253 (2012-12-12)
There exists a misunderstanding on the TAED-activated peroxide system in the textile industry that H(2)O(2) used in excess of the stoichiometric amount could produce an addition effect on bleaching of cotton under alkaline conditions. In this study, a critical reinvestigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service