Skip to Content
Merck
All Photos(1)

Key Documents

145610

Sigma-Aldrich

3-(Bromomethyl)benzonitrile

95%

Synonym(s):

α-Bromo-m-tolunitrile, 3-Cyanobenzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2C6H4CN
CAS Number:
Molecular Weight:
196.04
Beilstein:
742356
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

93-96 °C

functional group

bromo

SMILES string

BrCc1cccc(c1)C#N

InChI

1S/C8H6BrN/c9-5-7-2-1-3-8(4-7)6-10/h1-4H,5H2

InChI key

CVKOOKPNCVYHNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Bromomethyl)benzonitrile undergoes Suzuki cross-coupling reaction with bis(pinacolato)diboron.

Application

3-(Bromomethyl)benzonitrile was used in the synthesis of 3-(bromomethyl)benzaldehyde.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lydia C Gilday et al.
Dalton transactions (Cambridge, England : 2003), 41(23), 7092-7097 (2012-05-09)
Triazole and triazolium groups have been integrated into a zinc(II) metalloporphyrin-based structural framework to produce two porphyrin-cages for anion sensing applications. UV/visible spectroscopic titration investigations reveal both host systems exhibit strong anion binding affinities, with the positively-charged triazolium-porphyrin cage capable
Synthesis of benzylic boronates via palladium-catalyzed cross-coupling reaction of bis (pinacolato) diboron with benzylic halides.
Giroux A.
Tetrahedron Letters, 44(2), 233-235 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service